Skip to Content
Merck
All Photos(2)

Documents

810120C

Avanti

18:0 PEG2000 PE CF

Avanti Research - A Croda Brand 810120C

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[poly(ethylene glycol)2000-N′-carboxyfluorescein] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810120C-1mg)
pkg of 1 × 1 mL (810120C-500ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810120C

concentration

0.5 mg/mL (810120C-500ug)
1 mg/mL (810120C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[poly(ethylene glycol)2000-N′-carboxyfluorescein] (18:0 PEG2000 PE CF) lipid comprises hydrophilic polyethylene glycol (PEG) polymer in its head groups.

Application

18:0 PEG2000 PE CF may be used:
  • in in vitro release assays from nanoemulsion (NE)
  • for the labeling PEGylated LPD (liposome-polycation-DNA) for size exclusion chromatography studies
  • as a fluorescent probe to monitor shape transformation in giant vesicles

Biochem/physiol Actions

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[poly(ethylene glycol)2000-N′-carboxyfluorescein] (18:0 PEG2000 PE CF) conjugate is useful in designing biointerfaces. Lipid-anchored poly(ethylene glycol) (PEG) polymers aid in liposomal drug delivery. PEG-PE liposomes aid in the controlled release of drugs and are promising drug carriers. 18:0 PEG2000 PE CF, also called the DSPE−PEG2000, is a stable polymeric micelle, which favors hydrophobic drug circulation by preventing uptake by mononuclear phagocyte system. DSPE−PEG2000 is a potential drug carrier for cancer therapy.

Packaging

5 mL Clear Glass Sealed Ampule (810120C-1mg)
5 mL Clear Glass Sealed Ampule (810120C-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PEG-Lipid Post Insertion into Drug Delivery Liposomes Quantified at the Single Liposome Level
Eliasen R, et al.
Advanced Materials Interfaces, 6(9), 1801807-1801807 (2019)
Repeated injections of PEG-PE liposomes generate anti-PEG antibodies
Sroda K, et al.
Cellular & Molecular Biology Letters, 10, 37-47 (2005)
Nanoparticles evading the reticuloendothelial system: role of the supported bilayer
Li SD and Huang L
Biochimica et Biophysica Acta - Biomembranes, 1788(10), 2259-2266 (2009)
Molecular-matched materials for anticancer drug delivery and imaging
Wang D, et al.
Nanomedicine : nanotechnology, biology, and medicine, 10(19), 3003-3013 (2015)
Shape Transformations of Lipid Vesicles by Insertion of Bulky-Head Lipids
Tsuda S, et al.
PLoS ONE, 10(7), e0132963-e0132963 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service