Skip to Content
Merck
All Photos(1)

Key Documents

W527211

Sigma-Aldrich

Butyric anhydride

98%

Synonym(s):

Butanoic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CO)2O
CAS Number:
Molecular Weight:
158.19
Beilstein:
1099474
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

vapor density

5.45 (vs air)

vapor pressure

10 mmHg ( 79.5 °C)

Assay

98%

autoignition temp.

535 °F

expl. lim.

1.1 %, 104 °F
7.6 %, 144 °F

refractive index

n20/D 1.413 (lit.)

bp

198-199 °C (lit.)

mp

−75-−66 °C (lit.)

density

0.967 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

butter

SMILES string

CCCC(=O)OC(=O)CCC

InChI

1S/C8H14O3/c1-3-5-7(9)11-8(10)6-4-2/h3-6H2,1-2H3

InChI key

YHASWHZGWUONAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Butyric anhydride can be used to prepare various flavor & fragrance compounds like neryl butyrate, geranyl butyrate, and butyl butyrllactate.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Butyl butyrllactate, neryl butyrate, geranyl butyrate
Encyclopedia of Food and Color Additives, , year= (1997)
Butyl butyrllactate, neryl butyrate, geranyl butyrate
Encyclopedia of Food and Color Additives, , year= (1997)
Shiva Shanker Kaki et al.
Journal of oleo science, 64(8), 845-852 (2015-07-17)
Ferulic acid was modified to produce a novel phenolipid containing butyl chains. Ferulic acid was esterified with butanol to produce butyl ferulate which was further dihydroxylated followed by esterification with butyric anhydride to produce the phenolipid containing butyric acid. IR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service