Skip to Content
Merck
All Photos(3)

Key Documents

A88476

Sigma-Aldrich

4-Methoxybenzoyl chloride

99%

Synonym(s):

4-Anisoyl chloride, p-Anisoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4COCl
CAS Number:
Molecular Weight:
170.59
Beilstein:
471918
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.581 (lit.)

bp

262-263 °C (lit.)

mp

22 °C (lit.)

density

1.260 g/mL at 20 °C (lit.)

SMILES string

COc1ccc(cc1)C(Cl)=O

InChI

1S/C8H7ClO2/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3

InChI key

MXMOTZIXVICDSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methoxybenzoyl chloride is one of the reactive acylating agents that can react with carboxylic acids, alcohols and amines to yield respective carboxylic anhydrides, esters and amides.

4-methoxybenzoyl chloride reacts with potassium thiocyanate to yield isothiocyanate derivative via nucleophilic addition-elimination mechanism.

Application

  • 4-Methoxybenzoyl chloride can be used as radical precursor in visible-light photocatalysis to synthesize various heterocyclic compounds.
  • It can be used to synthesize acylphosphine ligands for the rhodium-catalyzed hydrosilylation of alkenes.
  • Incorporation of 4-methoxybenzoyl chloride modified indium tin oxide (ITO) as cathode for the fabrication of organic light-emitting diodes (OLEDs) has been reported.
  • 1,3 diketones synthesized from 4-methoxybenzoyl chloride can be used in one pot synthesis of various pyrazole derivatives.
  • It can also be used in the total synthesis of bioactive compounds like echinoside A and salinosporamide A.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of an acylphosphine ligand on the rhodium-catalyzed hydrosilylation of alkenes.
Li J, et al.
Journal of Organometallic Chemistry, 855, 7-11 (2018)
The modification of self-assembled monolayer on indium tin oxide as cathode in inverted bottom-emitting organic light-emitting diodes.
Xiong T and Ma D
Journal of Applied Physics, 104(6), 064506-064506 (2008)
1, 3-Diketones from acid chlorides and ketones: a rapid and general one-pot synthesis of pyrazoles.
Heller ST and Natarajan SR
Organic Letters, 8(13), 2675-2678 (2006)
Aroyl chlorides as novel acyl radical precursors via visible-light photoredox catalysis.
Xu S.M, et al.
Organic Chemistry Frontiers : An International Journal of Organic Chemistry / Royal Society of Chemistry, 4(7), 1331-1335 (2017)
Total synthesis of echinoside A, a representative triterpene glycoside of sea cucumbers.
Yu B, et al.
Angewandte Chemie (International Edition in English), 129(26), 7756-7760 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service