Skip to Content
Merck
All Photos(1)

Documents

W367303

Sigma-Aldrich

2-Ethylfuran

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O
CAS Number:
Molecular Weight:
96.13
FEMA Number:
3673
EC Number:
Council of Europe no.:
11706
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.092
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Halal
Kosher

Agency

meets purity specifications of JECFA

Assay

≥99%

refractive index

n20/D 1.439 (lit.)

bp

92-93 °C/768 mmHg (lit.)

density

0.912 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

coffee; ethereal; sweet

SMILES string

CCc1ccco1

InChI

1S/C6H8O/c1-2-6-4-3-5-7-6/h3-5H,2H2,1H3

InChI key

HLPIHRDZBHXTFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylfuran is one of the volatile components of bell pepper and certain fish species.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Comparison of natural polyphenol antioxidants from extra virgin olive oil with synthetic antioxidants in tuna lipids during thermal oxidation.
Medina I, et al.
Journal of Agricultural and Food Chemistry, 47(12), 4873-4879 (1999)
Study of the volatile compounds useful for the characterisation of fresh and frozen-thawed cultured gilthead sea bream fish by solid-phase microextraction gas chromatography?mass spectrometry.
Iglesias J, et al.
Food Chemistry, 115(4), 1473-1478 (2009)
Effect of tissue disruption on volatile constituents of bell peppers.
Wu CM & Liou SE.
Journal of Agricultural and Food Chemistry, 34(4), 770-772 (1986)
Static headspace gas chromatographic analyses to determine oxidation of fish muscle lipids during thermal processing.
Medina I, et al.
Journal of the American Oil Chemists' Society, 76(2), 231-236 (1999)
Francisco J Hidalgo et al.
Journal of agricultural and food chemistry, 62(49), 12045-12051 (2014-11-25)
The carbonyl-scavenging ability of 2-amino-1-methylbenzimidazole (AMBI) and the heterocyclic aromatic amine 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) was investigated in an attempt to identify new routes that can modify the carbonyl content of foods. The reaction of both AMBI and PhIP with 2-alkenals, 2,4-alkadienals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service