Skip to Content
Merck
All Photos(1)

Key Documents

W355305

Sigma-Aldrich

Isophorone

≥97%, FG

Synonym(s):

3,5,5-Trimethyl-2-cyclohexen-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O
CAS Number:
Molecular Weight:
138.21
FEMA Number:
3553
Beilstein:
1280721
EC Number:
Council of Europe no.:
4011
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.126
NACRES:
NA.21
grade:
FG
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012

vapor density

4.77 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

Assay

≥97%

autoignition temp.

864 °F

expl. lim.

3.8 %

refractive index

n20/D 1.476 (lit.)

bp

213-214 °C (lit.)

mp

−8 °C (lit.)

density

0.923 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; musty; cooling; camphoraceous; woody; fruity; sweet

SMILES string

CC1=CC(=O)CC(C)(C)C1

InChI

1S/C9H14O/c1-7-4-8(10)6-9(2,3)5-7/h4H,5-6H2,1-3H3

InChI key

HJOVHMDZYOCNQW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Odor at 1.0%
Taste at 30 ppm

Other Notes

Natural occurrence: Burley tobacco, cranberry, macadamia nuts, peas, roasted filbert, saffron, wine and osmanthus.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bojidarka B Ivanova
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 65(5), 1035-1040 (2006-07-11)
Possibilities of linear-dichroic infrared (IR-LD) spectroscopy based on oriented solid samples as suspension in nematic liquid crystal have been applied for detailed experimental IR-band assignment and structural information of 2-[5,5-dimethyl-3-(2-phenyl-vinil)-cyclohex-2-enylidene]-malononitrile, 2-[5,5-dimethyl-3-[2-(2-methoxyphenyl)vinyl]cyclo-hex-2-enylidene]malononitrile and 2-[3-[2-(2,4-dimethoxyphenyl)vinyl]-5,5-dimethylcyclohex-2-enylidene]malononitrile. The data of last two compounds have
Mourad Daoubi et al.
Journal of agricultural and food chemistry, 53(15), 6035-6039 (2005-07-21)
Diisophorone (1) was tested against two strains of the necrotrophic plant pathogen Botrytis cinerea. Fungal sensitivity varied according to the strain. B. cinera 2100 was more sensitive than B. cinereaUCA992: its mycelial growth was significantly inhibited at concentrations of 50
J L O'Donoghue et al.
Mutation research, 206(2), 149-161 (1988-10-01)
3 ketone solvents (methyl ethyl ketone (MEK), methyl isobutyl ketone (MiBK), and isophorone) were tested for potential genotoxicity. The assays of MEK and MiBK included the Salmonella/microsome (Ames) assay, L5178Y/TK+/- mouse lymphoma (ML) assay, BALB/3T3 cell transformation (CT) assay, unscheduled
Shilpa C Mhadgut et al.
Chemical communications (Cambridge, England), (8)(8), 984-985 (2004-04-08)
The sonochemical asymmetric hydrogenation of isophorone (3,3,5-trimethyl-2-cyclohexenone) by proline-modified Pd/Al2O3 catalysts is described; presonication of a commercial Pd/Al2O3-proline catalytic system resulted in highly enhanced enantioselectivities (up to 85% ee).
Errol V Mathias et al.
Langmuir : the ACS journal of surfaces and colloids, 24(3), 692-700 (2007-12-07)
Rf-IPDU-PEGs belong to a class of fluoroalkyl-ended poly(ethylene glycol) polymers (Rf-PEGs), where the IPDU (isophorone diurethane) functions as a linker to connect each end of the PEG chain to a fluoroalkyl group. The Rf-IPDU-PEGs form hydrogels in water with favorable

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service