Skip to Content
Merck
All Photos(1)

Documents

W291102

Sigma-Aldrich

Piperonal

≥99%, FCC, FG

Synonym(s):

1,3-Benzodioxole-5-carboxaldehyde, 3,4-(Methylenedioxy)benzaldehyde, Heliotropin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6O3
CAS Number:
Molecular Weight:
150.13
FEMA Number:
2911
Beilstein:
131691
EC Number:
Council of Europe no.:
104
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.016
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 182.60

vapor pressure

1 mmHg ( 87 °C)

Assay

≥99%

bp

264 °C (lit.)

mp

35-39 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

heliotropine

Organoleptic

cherry; sweet; vanilla

SMILES string

[H]C(=O)c1ccc2OCOc2c1

InChI

1S/C8H6O3/c9-4-6-1-2-7-8(3-6)11-5-10-7/h1-4H,5H2

InChI key

SATCULPHIDQDRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • The synthesis and characterisation of MDMA derived from a catalytic oxidation of material isolated from black pepper reveals potential route specific impurities.: This study explores the synthesis and characterization of MDMA from piperonal, highlighting potential impurities unique to this synthesis route. This research has implications for forensic science and the identification of synthetic routes for MDMA (Plummer et al., 2016).

  • Design, synthesis, and biological evaluation of platensimycin analogues with varying degrees of molecular complexity.: This paper details the synthesis of platensimycin analogues using piperonal derivatives. The study evaluates the biological activities of these analogues, contributing to the development of new antibacterial agents (Nicolaou et al., 2008).

  • Synthesis and use of 4-peptidylhydrazido-N-hexyl-1,8-naphthalimides as fluorogenic histochemical substrates for dipeptidyl peptidase IV and tripeptidyl peptidase I.: This research presents the synthesis of piperonal-based substrates for histochemical applications, enabling the study of enzyme activities in biochemical assays (Ivanov et al., 2009).

Biochem/physiol Actions

Taste at 10-50 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

150.1 °F

Flash Point(C)

65.62 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael S Macias et al.
Forensic science international, 195(1-3), 132-138 (2010-01-02)
Currently, in the field of odor detection, there is generally a wider variation in limit of detections (LODs) for canines than instruments. The study presented in this paper introduces an improved protocol for the creation of controlled odor mimic permeation
K C Nicolaou et al.
Journal of the American Chemical Society, 130(39), 13110-13119 (2008-09-06)
The molecular design, chemical synthesis, and biological evaluation of two distinct series of platensimycin analogues with varying degrees of complexity are described. The first series of compounds probes the biological importance of the benzoic acid subunit of the molecule, while
H Hatt et al.
Cellular and molecular biology (Noisy-le-Grand, France), 45(3), 285-291 (1999-07-01)
The human olfactory system can recognize and discriminate a large number of different odorant molecules. The detection of chemically distinct odorants begins with the binding of an odorant ligand to a specific receptor protein on the olfactory neuron cell surface.
W H Redd et al.
Journal of magnetic resonance imaging : JMRI, 4(4), 623-626 (1994-07-01)
Fifty-seven outpatients received either heliotropin (a vanillalike scent) with humidified air (n = 20) or humidified air alone (n = 37) via a nasal cannula during magnetic resonance imaging in the diagnostic workup for cancer. Delivery of heliotropin and air
S Peock et al.
Journal of the Royal Society of Health, 113(6), 292-294 (1993-12-01)
Piperonal, once used to kill lice in Australian hospitals, was acclaimed as an effective pediculicide (Corlette, 1925) by the standards of the day. It is unusual in also exhibiting a repellent action against lice, a property only recently realised. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service