Skip to Content
Merck
All Photos(1)

Key Documents

W278106

Sigma-Aldrich

γ-Nonanoic lactone

≥98%, FCC, FG

Synonym(s):

Aldehyde C18, Coconut aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O2
CAS Number:
Molecular Weight:
156.22
FEMA Number:
2781
EC Number:
Council of Europe no.:
178
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.001
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.447 (lit.)

bp

121-122 °C/6 mmHg (lit.)

density

0.976 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

coconut; creamy; fatty; oily; buttery; sweet

SMILES string

CCCCCC1CCC(=O)O1

InChI

1S/C9H16O2/c1-2-3-4-5-8-6-7-9(10)11-8/h8H,2-7H2,1H3

InChI key

OALYTRUKMRCXNH-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

γ-Nonanoic lactone is an oak lactone commonly found in wines aged in oak barrels. It is the main flavor component of coconut. γ-Nonanoic lactone is also a potential multi-species attractant lure for grain beetle pests.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The preparation and use of novel immobilised guanidine catalysts in base-catalysed epoxidation and condensation reactions.
Blanc AC, et al.
Green Chemistry, 2(6), 283-288 (2000)
Determination of volatile oak compounds in aged wines by multiple headspace solid-phase microextraction and gas chromatography?mass spectrometry (MHS-SPME?GC?MS).
Carrillo JD & Tena MT.
Analytical and Bioanalytical Chemistry, 385(5), 937-943 (2006)
Progress towards a multi-species lure: identification of components of food volatiles as attractants for three storage beetles.
Collins LE, et al.
Journal of Stored Products Research, 43(1), 53-63 (2007)
Laura E Korhonen et al.
Journal of medicinal chemistry, 48(11), 3808-3815 (2005-05-27)
The purpose of this study was to determine the cytochrome P450 1A2 (CYP1A2) inhibition potencies of structurally diverse compounds to create a comprehensive three-dimensional quantitative structure-activity relationship (3D-QSAR) model of CYP1A2 inhibitors and to use this model to predict the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service