Skip to Content
Merck
All Photos(1)

Documents

W271500

Sigma-Aldrich

Methyl laurate

≥98%, FG

Synonym(s):

Lauric acid methyl ester, Methyl dodecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CO2CH3
CAS Number:
Molecular Weight:
214.34
FEMA Number:
2715
Beilstein:
1767780
EC Number:
Council of Europe no.:
377
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.101
NACRES:
NA.21

biological source

synthetic

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.431 (lit.)

bp

262 °C/766 mmHg (lit.)

mp

4-5 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

coconut; creamy; fatty; soapy

SMILES string

CCCCCCCCCCCC(=O)OC

InChI

1S/C13H26O2/c1-3-4-5-6-7-8-9-10-11-12-13(14)15-2/h3-12H2,1-2H3

InChI key

UQDUPQYQJKYHQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl laurate is one of the key volatile flavor compounds in the avocado fruit. It is also reported to occur in Polygonum bistorta L. (snakeroot) and γ-aminobutyric acid (GABA) tea.

Application


  • Frustrated Lewis pair catalyst realizes efficient green diesel production.: Methyl laurate is employed as a key reactant in the development of an efficient and environmentally friendly catalyst system for the production of green diesel. This breakthrough offers a sustainable alternative to traditional diesel production methods (Li et al., 2024).

  • Sodium phosphate solid base catalysts for production of novel biodiesel by transesterification reaction.: This paper explores the use of sodium phosphate solid base catalysts in the transesterification of methyl laurate to produce biodiesel. The research demonstrates high efficiency and novel catalyst design, contributing to advancements in sustainable fuel production (Zhao et al., 2023).

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

282.2 °F - closed cup

Flash Point(C)

139 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of the volatile fraction of Polygonum bistorta L. at different growing stages and evaluation of its antimicrobial activity against two major honeybee (Apis mellifera) pathogens.
Cecotti R, et al.
Chemistry and Biodiversity, 9(2), 359-369 (2012)
Identification of volatile compounds in cultivars barker, collinson, fortuna and geada of avocado (Persea americana, Mill.) fruit.
Galvao MDS, et al.
Food Sci. Technol., 36(3), 439-447 (2016)
Study on flavour volatiles of ?-aminobutyric acid (GABA) green tea.
Zeng Z, et al.
African Journal of Biotechnology, 11(51), 11333-11341 (2012)
Y Komata et al.
Biological & pharmaceutical bulletin, 17(5), 705-708 (1994-05-01)
The effect of long chain fatty acid (FA) and its analogs on the accumulation of thiamine disulfide (TDS) in rat skin using propylene glycol as a vehicle was studied in vitro. Lauric acid (12:0) increased the accumulation of TDS in
D A Dimas et al.
AAPS PharmSciTech, 1(2), E16-E16 (2004-01-20)
The effects of coating thickness, type of adhesive, and type and concentration of enhancer on the mechanical properties of two acrylic pressure-sensitive adhesives (PSAs) were investigated using a 2(4) factorial design and an optimization technique. Sixteen formulations containing 0% or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service