Skip to Content

1424233

USP

Methyl caprate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Methyl decanoate, Capric acid methyl ester, Decanoic acid methyl ester, Methyl caprate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)8COOCH3
CAS Number:
Molecular Weight:
186.29
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1759170
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

methyl caprate

manufacturer/tradename

USP

refractive index

n20/D 1.425 (lit.)

bp

108 °C/10 mmHg, 224 °C (lit.)

mp

−14-−11 °C (lit.)

density

0.871 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCCCC(=O)OC

InChI

1S/C11H22O2/c1-3-4-5-6-7-8-9-10-11(12)13-2/h3-10H2,1-2H3

InChI key

YRHYCMZPEVDGFQ-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methyl caprate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Glyceryl Monocaprylate
  • Glyceryl Mono and Dicaprylate
  • Glyceryl Mono and Dicaprylocaprate
  • Glyceryl Monocaprylocaprate
  • Glyceryl Tricaprylate
  • Saw Palmetto Capsules
  • Powdered Saw Palmetto
  • Saw Palmetto

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Methyl caprate


pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library