Skip to Content
Merck
All Photos(3)

Key Documents

L2602

Sigma-Aldrich

Linalool

97%

Synonym(s):

(±)-3,7-Dimethyl-1,6-octadien-3-ol, (±)-3,7-Dimethyl-3-hydroxy-1,6-octadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)(OH)CH=CH2
CAS Number:
Molecular Weight:
154.25
Beilstein:
1721488
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.17 mmHg ( 25 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.462 (lit.)

bp

194-197 °C/720 mmHg (lit.)

density

0.87 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C\CCC(C)(O)C=C

InChI

1S/C10H18O/c1-5-10(4,11)8-6-7-9(2)3/h5,7,11H,1,6,8H2,2-4H3

InChI key

CDOSHBSSFJOMGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Linalool is generally used as a starting material in the total synthesis of agelasine E analogs[1] and vitamin E derivatives.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

171.0 °F - Pensky-Martens closed cup

Flash Point(C)

77.2 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and antimycobacterial activity of agelasine E and analogs.
Bakkestuen AK, et al.
Organic & Biomolecular Chemistry, 3(6), 1025-1033 (2005)
A convenient access to (all-rac)-α-tocopherol acetate from linalool and dihydromyrcene
Gembus V, et al.
Bulletin of the Chemical Society of Japan, 82(7), 829-842 (2009)
Ali M Al Hazmi et al.
Organic letters, 16(19), 5104-5107 (2014-09-17)
The permanganate-mediated oxidative cyclization of a series of 2-methylenehept-5-eneoates bearing different chiral auxiliaries was investigated, leading to the discovery of trans-2-tritylcyclohexanol (TTC) as a highly effective chiral controller for the formation of the 2,5-substituted THF diol product with high diastereoselectivity
Meixia Huo et al.
The Journal of surgical research, 180(1), e47-e54 (2012-12-12)
Inflammation, characterized by redness, swelling, pain and a sensation of heat, is one of the body's self-defense systems. Although the inflammation response has an important role in host survival, it also leads to chronic inflammatory diseases. Linalool is a natural
A toxicologic and dermatologic assessment of linalool and related esters when used as fragrance ingredients.
D Bickers et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(7), 919-942 (2003-06-14)

Questions

1–5 of 5 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. I had order it previously and I wanted to do gcms of it. For that, I need to know what is it dissolved in?

    1 answer
    1. This product is a neat or pure liquid and has not been diluted in a solvent. This material has a molecular weight of 154.25 daltons and a density of 0.87 g/mL at 25 °C.

      Helpful?

  4. Hi.. for what application it can be used ??

    1 answer
    1. This product is intended for use in general laboratory research. As listed in the DESCRIPTION section under 'Application', Linalool may be used as a starting material in the total synthesis of agelasine E analogs[1] and vitamin E derivatives. Additional application areas may be accessed in the Peer Reveiwed Papers related to this compound. Please see the link below to access these publications:
      https://www.sigmaaldrich.com/search/l2602?focus=papers&page=1&perpage=30&sort=relevance&term=L2602&type=citation_search

      Helpful?

  5. Is linalool is liquid or solid in nature if it is liquid 5g of linalool contain how much ml of linalool

    1 answer
    1. The reported density of this material is 0.87 g/mL. The volume of linalool offered in the 5g package size is approximately 5.75mL.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service