Skip to Content
Merck
All Photos(4)

Key Documents

H40807

Sigma-Aldrich

5-Hydroxymethyl-2-furaldehyde

99%

Synonym(s):

5-(Hydroxymethyl)furfural, 5-Hydroxymethyl-2-furancarboxaldehyde, HMF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
Beilstein:
110889
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.562 (lit.)

bp

114-116 °C/1 mmHg (lit.)

mp

28-34 °C (lit.)

density

1.243 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccc(CO)o1

InChI

1S/C6H6O3/c7-3-5-1-2-6(4-8)9-5/h1-3,8H,4H2

InChI key

NOEGNKMFWQHSLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Hydroxymethyl-2-furaldehyde (HMF) can be used in the synthesis of:
  • Furan-2,5-dicarboxaldehyde (FDA) using a vanadyl phosphate-based catalyst via selective oxidation.
  • Prepolymer and antiviral precursor, 5,5′-oxy(bismethylene)-2-furaldehyde (OBMF) using solid acid catalysts via etherification reaction.
  • Diformylfurane (DFF) by aerobic oxidation using pyridine-vanadyl complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective oxidation of 5-hydroxymethyl-2-furaldehyde to furan-2, 5-dicarboxaldehyde by catalytic systems based on vanadyl phosphate.
Carlini C, et al.
Applied Catalysis A: General, 289(2), 197-204 (2005)
Chemicals from biomass: Aerobic oxidation of 5-hydroxymethyl-2-furaldehyde into diformylfurane catalyzed by immobilized vanadyl-pyridine complexes on polymeric and organofunctionalized mesoporous supports.
Navarro OC, et al.
Topics in Catalysis, 52(3), 304-314 (2009)
Chemicals from biomass: Etherification of 5-hydroxymethyl-2-furfural (HMF) into 5, 5?(oxy-bis (methylene)) bis-2-furfural (OBMF) with solid catalysts.
Casanova O, et al.
J. Catal., 275(2), 236-242 (2010)
Yin-Ying Lee et al.
Physical chemistry chemical physics : PCCP, 14(40), 13914-13917 (2012-09-20)
Mesoporous silica nanoparticles functionalized with both sulfonic acid (HSO(3)) and ionic liquid (ILs) were synthesized and applied as effective and recyclable catalysts for generating 5-hydroxymethylfurfural (HMF) from fructose. For the first time a high HMF yield of 72.5% was achieved
Monika Krystof et al.
ChemSusChem, 6(4), 630-634 (2013-03-05)
5-Hydroxymethylfurfural (HMF) is a valuable biomass-derived building block. Among possible HMF valorization products, a broad range of HMF esters can be synthesized. These HMF esters have found some promising applications, such as monomers, fuels, additives, surfactants, and fungicides, and thus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service