Skip to Content
Merck
All Photos(1)

Key Documents

W238600

Sigma-Aldrich

1,4-Dimethoxybenzene

99%, FG

Synonym(s):

p-Methoxyanisole, Dimethylhydroquinone, Hydroquinone dimethyl ether, NSC 7483

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OCH3)2
CAS Number:
Molecular Weight:
138.16
FEMA Number:
2386
Beilstein:
774605
EC Number:
Council of Europe no.:
2059
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.034
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

vapor density

4.8 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

99%

autoignition temp.

1463 °F

bp

213 °C (lit.)

mp

54-56 °C (lit.)

density

1.053 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fennel; green; sweet

SMILES string

COc1ccc(OC)cc1

InChI

1S/C8H10O2/c1-9-7-3-5-8(10-2)6-4-7/h3-6H,1-2H3

InChI key

OHBQPCCCRFSCAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Dimethoxybenzene can be used as a flavoring agent in the food industry. It s one of the main volatile constituents identified inTuber brumale, Vietnamese green tea and lotus tea.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavor components in Vietnamese green tea and lotus tea.
Nguyen TT & Yamanishi T
Agricultural and Biological Chemistry, 39(6), 1263-1267 (1975)
Determination of volatile organic compounds from truffles via SPME-GC-MS
Journal of Chromatographic Science, 42(6), 299-305 (2004)
Burdock, GA
Encyclopedia of Food and Color Additives, 1, 822-823 (1997)
Burdock, GA
Encyclopedia of Food and Color Additives, 1, 822-823 (1997)
D W Kim et al.
Organic letters, 3(3), 445-447 (2001-06-29)
[figure: see text] Fused 1,4-dimethoxybenzenes could be oxidized to benzoquinones by either direct oxidation or demethylation-oxidation. The oxidative demethylation of 5,8-dimethoxy-2-methylquinoline using 1.1 equiv of NBS in aqueous THF and a catalytic amount of H2SO4 at 20 degrees C for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service