Skip to Content
Merck
All Photos(3)

Key Documents

W217816

Sigma-Aldrich

Butyl alcohol

greener alternative

natural, ≥99.5%, FCC, FG

Synonym(s):

1-Butanol, n-Butanol, Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12
FEMA Number:
2178
Beilstein:
969148
EC Number:
Council of Europe no.:
52c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.004
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

FG
Halal
Kosher
natural

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

vapor density

2.55 (vs air)

Assay

≥99.5%

form

liquid

autoignition temp.

649 °F

expl. lim.

11.2 %

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.399 (lit.)

pH

7 (20 °C, 70 g/L)

bp

116-118 °C (lit.)

mp

−90 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

balsamic; sweet; whiskey

SMILES string

CCCCO

InChI

1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

LRHPLDYGYMQRHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl alcohol is an aliphatic alcohol that can be used as a flavoring agent.[1] It has been identified as one of the main volatile components of mushrooms.[2]
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • A new HS-SPME-GC-MS analytical method to identify and quantify compounds responsible for changes in the volatile profile in five types of meat products during aerobic storage at 4 °C.: This paper presents a novel analytical method to detect and quantify volatile compounds, such as butyl alcohol, that affect the quality and safety of stored meat products (Acquaticci et al., 2024).

  • Impact of non-Saccharomyces yeasts derived from traditional fermented foods on beer aroma: Analysis based on HS-SPME-GC/MS combined with chemometrics.: This research explores how non-Saccharomyces yeasts influence beer aroma profiles, with a focus on the role of butyl alcohol in the fermentation process and flavor development (Cao et al., 2024).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - Pensky-Martens closed cup

Flash Point(C)

35 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA
Encyclopedia of Food and Color Additives, 1, 319-320 (1997)
Volatile components of mushroom (Agaricus subrufecens)
Chen CC & Wu CM
Journal of Food Science, 49(4), 1208-1209 (1984)
R W Wulkan et al.
The International journal of biochemistry, 18(11), 1045-1051 (1986-01-01)
The butanol extraction method of Morton (1950), a routine step in enzyme purification, is discussed with special reference to a hydrophobic form of alkaline phosphatase from human liver tissue. This form slowly precipitates from butanol-extracted liver tissue homogenates stored at
Joel G Davis et al.
Nature, 491(7425), 582-585 (2012-11-23)
Hydrophobic hydration is considered to have a key role in biological processes ranging from membrane formation to protein folding and ligand binding. Historically, hydrophobic hydration shells were thought to resemble solid clathrate hydrates, with solutes surrounded by polyhedral cages composed
Pazhamalai Anbarasan et al.
Nature, 491(7423), 235-239 (2012-11-09)
Nearly one hundred years ago, the fermentative production of acetone by Clostridium acetobutylicum provided a crucial alternative source of this solvent for manufacture of the explosive cordite. Today there is a resurgence of interest in solventogenic Clostridium species to produce

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service