Skip to Content
Merck
All Photos(3)

Key Documents

W205710

Sigma-Aldrich

Isoamyl alcohol

greener alternative

natural, ≥98%, FG

Synonym(s):

3-Methyl-1-butanol, Isoamyl alcohol, Isopentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2OH
CAS Number:
Molecular Weight:
88.15
FEMA Number:
2057
Beilstein:
1718835
EC Number:
Council of Europe no.:
51
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.003
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 20 °C)

Assay

≥98%

form

liquid

autoignition temp.

644 °F

expl. lim.

1.2-9 %, 100 °F

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

refractive index

n20/D 1.406 (lit.)

pH

5.6 (20 °C, 25 g/L)

bp

130 °C (lit.)

mp

−117 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

banana; oily; fruity; whiskey

SMILES string

CC(C)CCO

InChI

1S/C5H12O/c1-5(2)3-4-6/h5-6H,3-4H2,1-2H3

InChI key

PHTQWCKDNZKARW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Isoamyl alcohol is a cyanobacterial volatile organic compound that has been identified as one of the main aroma/flavor constituents in banana fruit, karanda fruit and in whey spirit.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”. Click here to view its Biobased natural statement.

Application


  • Gradually supply of isoamyl alcohol increases the isoamyl acetate production in solid-state fermentation.: This study demonstrates how controlled addition of Isoamyl alcohol enhances isoamyl acetate production in solid-state fermentation, suggesting applications in industrial biotechnology for flavor enhancement (Sainz-Mellado et al., 2023 link).

  • Key aroma compounds and metabolic profiling of Debaryomyces hansenii L1-1-fermented Flos Sophorae.: Investigation into the metabolic pathways in Debaryomyces hansenii fermentation, where Isoamyl alcohol serves as a crucial intermediate, contributing to the unique aroma profile of the fermented product (Zhang et al., 2021 link).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

110.3 °F - closed cup

Flash Point(C)

43.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L-leucine as a precursor of isoamyl alcohol and isoamyl acetate, volatile aroma constituents of banana fruit discs
Myers MJ, et al.
Phytochemistry, 9(8), 1693-1700 (1970)
Characterisation of volatile compounds in an alcoholic beverage produced by whey fermentation.
Dragone G, et al.
Food Chemistry, 112(4), 929-935 (2009)
Analytical aspects of cyanobacterial volatile organic compounds for investigation of their production behavior.
Fujise D, et al
Journal of Chromatography A, 1217(39), 6122-6125 (2010)
Volatile flavor constituents of Karanda (Carissa carandas L.) fruit.
Pino JA, et al.
J. Essent. Oil Res., 16(5), 432-434 (2004)
Mary J Dunlop et al.
Molecular systems biology, 7, 487-487 (2011-05-11)
Many compounds being considered as candidates for advanced biofuels are toxic to microorganisms. This introduces an undesirable trade-off when engineering metabolic pathways for biofuel production because the engineered microbes must balance production against survival. Cellular export systems, such as efflux

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service