Skip to Content
Merck
All Photos(1)

Key Documents

T69000

Sigma-Aldrich

3,4,5-Trimethoxybenzoic acid

ReagentPlus®, 99%

Synonym(s):

Gallic acid trimethyl ether, Trimethylgallic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CO2H
CAS Number:
Molecular Weight:
212.20
Beilstein:
884655
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

product line

ReagentPlus®

Assay

99%

bp

225-227 °C/10 mmHg (lit.)

mp

168-171 °C (lit.)

SMILES string

COc1cc(cc(OC)c1OC)C(O)=O

InChI

1S/C10H12O5/c1-13-7-4-6(10(11)12)5-8(14-2)9(7)15-3/h4-5H,1-3H3,(H,11,12)

InChI key

SJSOFNCYXJUNBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Tateno et al.
Mutation research, 327(1-2), 237-246 (1995-03-01)
Many inhibitors of tubulin polymerization have a trimethoxybenzene ring in their molecules. Such trimethoxybenzoic compounds and their analogues may therefore have a potency to induce meiotic nondisjunction of oocytes. In this study, a single dose of reserpine (0.5 microgram/g body
S Kleis-San Francisco et al.
The Journal of experimental zoology, 244(1), 133-143 (1987-10-01)
In the amphibian ovarian follicle, progesterone production is thought to induce maturation of the enclosed oocyte. Intracellular mechanisms regulating these events in the somatic and germ cells are incompletely understood. However, calcium appears to play a role in the production
[Synthesis and the study of the effect of new 3,4,5-trimethoxybenzoic acid derivatives on the central nervous system].
R Glinka et al.
Acta poloniae pharmaceutica, 42(2), 117-122 (1985-01-01)
Yan-Jun Hu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 65(3-4), 988-992 (2006-05-09)
The interaction between 3,4,5-trimethoxybenzoic acid (TMBA) and bovine serum albumin (BSA) was studied by fluorescence and UV-vis absorption spectroscopy. In the mechanism discussion, it was proved that the fluorescence quenching of BSA by TMBA is a result of the formation
H Naviasky
Journal of pharmaceutical sciences, 73(4), 542-545 (1984-04-01)
An ion-pair column chromatographic/UV spectrophotometric method for assaying trimethobenzamide hydrochloride in capsules and injections is presented, as well as a method for the detection of 3,4,5- trimethoxybenzoic acid in trimethobenzamide hydrochloride bulk drug and dosage forms. Results obtained by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service