Skip to Content
Merck
All Photos(1)

Key Documents

D132004

Sigma-Aldrich

3,5-Dimethoxybenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CO2H
CAS Number:
Molecular Weight:
182.17
Beilstein:
511834
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

178-180 °C (lit.)

SMILES string

COc1cc(OC)cc(c1)C(O)=O

InChI

1S/C9H10O4/c1-12-7-3-6(9(10)11)4-8(5-7)13-2/h3-5H,1-2H3,(H,10,11)

InChI key

IWPZKOJSYQZABD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Dimethoxybenzoic acid (3,5-DmeoxBA) can be used as a reactant for the synthesis of:
  • 5,7-Dimethoxy-3,4-diphenylisocoumarin by coupling with diphenylacetylene.
  • Biotin dimedone, a reagent used in the study of protein sulfenation.

It can also be used as a ligand to synthesize lanthanide complexes [Ln(3,5-DmeoxBA)3(phen)]2; where phen is 1,10-phenanthroline.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodium-and iridium-catalyzed oxidative coupling of benzoic acids with alkynes via regioselective C- H bond cleavage.
Ueura K, et al.
The Journal of Organic Chemistry, 72(14), 5362-5367 (2007)
Crystal structures, luminescence, and thermodynamic properties of lanthanide complexes with 3, 5-dimethoxybenzoic acid and 1, 10-phenanthroline.
Zheng JR, et al.
The Journal of Chemical Thermodynamics, 57(9), 169-177 (2013)
Protein sulfenation as a redox sensor: proteomics studies using a novel biotinylated dimedone analogue.
Charles RL, et al.
Molecular and Cellular Proteomics, 6(9), 1473-1484 (2007)
Marta Olech et al.
Molecules (Basel, Switzerland), 25(8) (2020-04-25)
Fast and reliable determination of polyphenols is a quite common goal during investigation of new plant materials and herbal products, their standardization, quality control, or chemo-taxonomical studies. The aim of this study was to develop and validate methods based on
Alla V Lipeeva et al.
European journal of medicinal chemistry, 100, 119-128 (2015-06-17)
A series of 2-(4-R-triazolyl)substituted 3-oxo-2,3-dihydrofurocoumarins have been synthesized by a regioselective cycloaddition of 2-azidooreoselone 1 or 2-azido-9-[(4-methylpiperazin-1-yl)methyl]oreoselone 2 with various alkynes in the presence of Cu(II)/ascorbate in water/methylene chloride reaction medium. The structure of 2-azidooreoselone was established by X-ray structure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service