Skip to Content
Merck
All Photos(3)

Key Documents

T15156

Sigma-Aldrich

3,4,5,6-Tetrahydro-2-pyrimidinethiol

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N2S
CAS Number:
Molecular Weight:
116.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

mp

210-212 °C (lit.)

SMILES string

S=C1NCCCN1

InChI

1S/C4H8N2S/c7-4-5-2-1-3-6-4/h1-3H2,(H2,5,6,7)

InChI key

NVHNGVXBCWYLFA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A capillary gas-liquid chromatographic method for determination of ethylenethiourea and prophylenethiourea in hops, beer, and grapes.
S Nitz et al.
Journal of agricultural and food chemistry, 30(3), 593-596 (1982-05-01)
James R Startin et al.
Food additives and contaminants, 22(3), 245-250 (2005-07-16)
A robust and sensitive method for the determination of ethylenethiourea (ETU) and iso-propylenethiourea (i-PTU) in foods is reported. ETU and i-PTU were extracted by blending with dichloromethane (DCM) in the presence of sodium sulphate, sodium carbonate, thiourea and ascorbic acid.
Gas-liquid chromatographic method for determining propylenthiourea in rat tissues and fluids.
S Lembo et al.
Journal of chromatography, 267(2), 427-430 (1983-09-16)
M Angela Montesano et al.
Journal of exposure science & environmental epidemiology, 17(4), 321-330 (2007-04-19)
Because of increasing concern about widespread use of insecticides and fungicides, we have developed a highly sensitive analytical method to quantify urine-specific urinary biomarkers of the organophosphorus pesticides acephate, methamidophos, omethoate, dimethoate, and two metabolites from the fungicides alkylenebis-(dithiocarbamate) family:
A Rolandi et al.
Mutation research, 135(3), 193-197 (1984-03-01)
The possible clastogenic activity of Propineb, Propineb technical grade and of its main metabolite, propylene-thiourea (PLTU), was investigated by the micronucleus test in mice according to Schmid. No statistically significant increase in the percentage of micronuclei was observed at any

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service