All Photos(4)
About This Item
Linear Formula:
HOOC(CH2)6COOH
CAS Number:
Molecular Weight:
174.19
Beilstein:
1210161
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
bp
230 °C/15 mmHg (lit.)
mp
139-146 °C
140-144 °C (lit.)
SMILES string
OC(=O)CCCCCCC(O)=O
InChI
1S/C8H14O4/c9-7(10)5-3-1-2-4-6-8(11)12/h1-6H2,(H,9,10)(H,11,12)
InChI key
TYFQFVWCELRYAO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Suberic acid can be used to synthesize:
- Perfluorinated analogs of suberoyl anilide hydroxamic acid (SAHA) as potent histone deacetylase inhibitors.
- Hemiester analogs of pregnenolone sulfate (PES).
- Cellulose acetate suberate, a cellulose ω-carboxyalkanoate.
- It can also act as a cross-linker for poly(vinyl alcohol) (PVA). The cross-linked molecule shows enhanced tensile strength when compared to neat PVA.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
410.0 °F - closed cup
Flash Point(C)
210 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Crosslinking of Polyvinyl Alcohol (PVA) and Effect of Crosslinker Shape (Aliphatic and Aromatic) Thereof.
Sonker A K, et al.
Journal of Polymers and the Environment, 26(5), 1782-1794 (2018)
Positive Modulators of the N-Methyl-d-aspartate Receptor: Structure?Activity Relationship Study of Steroidal 3-Hemiesters.
Krausova B, et al.
Journal of Medicinal Chemistry, 61(10), 4505-4516 (2018)
Perfluorinated HDAC inhibitors as selective anticancer agents.
Walton J W, et al.
Organic & Biomolecular Chemistry, 15(43), 9186-9190 (2017)
Rifampin stability and solution concentration enhancement through amorphous solid dispersion in cellulose ω-carboxyalkanoate matrices.
Arca H C, et al.
Journal of Pharmaceutical Sciences, 107(1), 127-138 (2018)
S Hayakawa
Zeitschrift fur allgemeine Mikrobiologie, 22(5), 309-326 (1982-01-01)
Through the isolation and identification of a wide variety of degradation products formed from bile acids by microorganisms, a unified scheme for the complete degradation of bile acids to carbon dioxide and water has been proposed and discussed. The proposed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service