Skip to Content
Merck
All Photos(3)

Key Documents

D1009

Sigma-Aldrich

Dodecanedioic acid

99%

Synonym(s):

Decane-1,10-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CH2)10COOH
CAS Number:
Molecular Weight:
230.30
Beilstein:
1782580
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

21 mmHg ( 222 °C)

Assay

99%

form

pellets

bp

245 °C/10 mmHg (lit.)

mp

127-129 °C (lit.)

SMILES string

OC(=O)CCCCCCCCCCC(O)=O

InChI

1S/C12H22O4/c13-11(14)9-7-5-3-1-2-4-6-8-10-12(15)16/h1-10H2,(H,13,14)(H,15,16)

InChI key

TVIDDXQYHWJXFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dodecanedioic acid (DDA) is a medium-chaindicarboxylic fatty acid commonly used in the production of polyamidenylon-6,12. This material is recognized for its resistance to heat andchemicals, making it suitable for various technical applications, such as heatand chemical-resistant sheaths.

Application

Dodecanedioic acid can be used as a reactant in the:
  • Solvent-free synthesis of biobased polyester polyols by reacting with 1,4-pentanediol.
  • Arylboronic acid-catalyzed direct condensation with ureas to synthesize N,N′-diacyl-2-imidazolidones.
It can also be used as a catalyst for the allylation of benzoquinones to synthesize lapachol using allyl alcohols or acetates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

428.0 °F - closed cup

Flash Point(C)

220 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harsha Ramaraju et al.
PloS one, 15(2), e0229112-e0229112 (2020-02-23)
Development of biodegradable shape memory elastomers (SMEs) is driven by the growing need for materials to address soft tissue pathology using a minimally invasive surgical approach. Composition, chain length and crosslinking of biocompatible polymers like PCL and PLA have been
Susanne Ditlevsen et al.
Bulletin of mathematical biology, 67(3), 547-561 (2005-04-12)
Deterministic and stochastic differential equations models of the uptake of dodecanedioic acid (C12) are fitted to experimental data obtained on nine isolated, perfused rat livers. 11500 mug of C12 were injected as a bolus into the perfusing liver solution. The
Christopher J van der Gast et al.
Environmental microbiology, 6(3), 254-263 (2004-02-12)
In order for established bioreactors to be effective for treating chemically mixed wastes such as metal working fluids (MWF) it is essential that they harbour microbial populations that can maintain sufficient active biomass and degrade each of the chemical constituents
Lieven Van Meulebroek et al.
Analytical chemistry, 89(22), 12502-12510 (2017-10-21)
As lipids are assigned a plethora of biological functions, it is evident that dysregulated lipid metabolism signifies a key element in many pathological conditions. With this rationale, this study presents a validated lipidomics platform to map the fecal lipidome, which
Stanley H Korman et al.
Molecular genetics and metabolism, 86(3), 337-343 (2005-09-09)
Detection of hepatic carnitine palmitoyltransferase I (CPT IA) deficiency by metabolite screening may be problematic. The urine organic acid profile is generally said to be normal and no abnormal or increased acylcarnitine species are evident on bloodspot tandem MS examination.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service