Skip to Content
Merck
All Photos(2)

Key Documents

R2206

Sigma-Aldrich

Rubrene

powder

Synonym(s):

5,6,11,12-Tetraphenylnaphthacene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H28
CAS Number:
Molecular Weight:
532.67
Beilstein:
1917339
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder

mp

330-335 °C (lit.)

λmax

299 nm

OLED Device Performance

ITO/CuPc/NPD/Alq3:Rubrene (5%): DCM2 (2%)/Alq3/Mg:In

  • Color: red
  • Max. Luminance: 7780 Cd/m2

ITO/PEDOT:PSS/EHCz:Rubrene (1 wt%)/Cs2CO3:ITO
  • Color: red
  • Max. EQE: 0.03 %

SMILES string

c1ccc(cc1)-c2c3ccccc3c(-c4ccccc4)c5c(-c6ccccc6)c7ccccc7c(-c8ccccc8)c25

InChI

1S/C42H28/c1-5-17-29(18-6-1)37-33-25-13-14-26-34(33)39(31-21-9-3-10-22-31)42-40(32-23-11-4-12-24-32)36-28-16-15-27-35(36)38(41(37)42)30-19-7-2-8-20-30/h1-28H

InChI key

YYMBJDOZVAITBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for chemiluminescence research and for transition metal complex ligation.
Reagent for chemiluminescence research[1] and for transition metal complex ligation.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inorganic Chemistry, 32, 1078-1078 (1993)
Rui M Pinto et al.
Journal of the American Chemical Society, 137(22), 7104-7110 (2015-05-21)
Exciton diffusion is at the heart of most organic optoelectronic devices' operation, and it is currently the most limiting factor to their achieving high efficiency. It is deeply related to molecular organization, as it depends on intermolecular distances and orbital
Marina Pivetta et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 11(7), 1558-1569 (2010-04-22)
Coverage-dependent self-assembly of rubrene molecules on different noble metal surfaces, Au(111) and Au(100), Ag(111) and Ag(100), is presented. On Au(111), the homochiral supramolecular assemblies evolve with increasing rubrene coverage from very small structures composed of a few molecules, to honeycomb
Surface potential mapping of SAM-functionalized organic semiconductors by Kelvin probe force microscopy.
David J Ellison et al.
Advanced materials (Deerfield Beach, Fla.), 23(4), 502-507 (2011-01-22)
Motonobu Murakami et al.
The journal of physical chemistry. A, 112(4), 635-642 (2008-01-05)
The investigations were made on photoinduced electron transfer (ET) from the singlet excited state of rubrene (1RU*) to p-benzoquinone derivatives (duroquinone, 2,5-dimethyl-p-benzoquinone, p-benzoquinone, 2,5-dichloro-p-benzoquinone, and p-chloranil) in benzonitrile (PhCN) by using the steady state and time-resolved spectroscopies. The photoinduced ET

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service