Skip to Content
Merck
All Photos(2)

Key Documents

P35316

Sigma-Aldrich

Diphenyl sulfide

98%

Synonym(s):

Phenyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2S
CAS Number:
Molecular Weight:
186.27
Beilstein:
1907932
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.6327 (lit.)

bp

296 °C (lit.)

mp

−40 °C (lit.)

density

1.113 g/mL at 20 °C (lit.)

SMILES string

S(c1ccccc1)c2ccccc2

InChI

1S/C12H10S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10H

InChI key

LTYMSROWYAPPGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

X Yang et al.
Guang pu xue yu guang pu fen xi = Guang pu, 21(4), 475-477 (2003-08-30)
Substitution of diphenyl sulfurine to dodecarbonyl-triruthenium and dodecarbonyl-triiron were studied respectively. Infrared spectra were used in characterization of the reactions and to follow the reaction processes from starting to finishing. Spectra data showed that the new compounds appeared and starting
Jiaqi Shi et al.
Chemosphere, 88(7), 844-854 (2012-05-02)
Twenty-nine congeners of polychlorinated diphenyl sulfide (PCDPS) theoretically possible were synthesized and purified, and their octanol-water partition coefficients (logK(ow)) and high performance liquid chromatography capacity factors (HPLC-logk') were determined. Then the constitutional and structural descriptors were obtained respectively to establish
Xuesheng Zhang et al.
Environmental toxicology and chemistry, 31(7), 1485-1493 (2012-05-03)
Polychlorinated diphenyl sulfides (PCDPSs), a series of dioxin-like compounds, have been detected in various environmental samples. However, information on the toxicity of these compounds is limited. In the present study, the toxic effects of PCDPSs were assessed after acute and
Julia Wellsow et al.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques, 5(3), 245-257 (2003-01-30)
Imaging the serotonin transporter (SERT) with Positron Emission Tomography (PET) provides a useful tool for understanding alterations of the serotonergic system. However, no optimal PET radiotracer for the SERT yet exists. The main purpose of this study was to design
Ruijuan Qu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 81(1), 261-269 (2011-07-15)
Tetrachlorinated diphenyl sulfides (TCDPSs) are environmentally interesting compounds. In this paper, both experimental and theoretical studies on IR and (1)H NMR as well as (13)C NMR chemical shifts of 4 synthesized TCDPSs have been carried out. The optimized geometries were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service