Skip to Content
Merck
All Photos(2)

Key Documents

GF74432392

Cobalt

wire reel, 0.1m, diameter 0.05mm, hard, 99.9%

Synonym(s):

Cobalt, CO005080

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
MDL number:
UNSPSC Code:
12141710
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 744-323-92

resistivity

6.24 μΩ-cm, 20°C

L × diam.

0.1 m × 0.05 mm

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth M Unice et al.
Chemico-biological interactions, 216, 53-74 (2014-04-15)
An updated biokinetic model for human exposures to cobalt (Co) was developed based on a comprehensive set of human pharmacokinetics data collected from five male and five female volunteers who ingested ∼1 mg Co/day of a Co supplement for 3
K Praveena et al.
Journal of nanoscience and nanotechnology, 14(6), 4371-4376 (2014-04-18)
The Cobalt ferrite (CoFe2O4) powders were synthesized by Co-precipitation method. The as prepared ferrite powders were incorporated into a polyaniline matrix at various volumetric ratios. The as prepared composites of ferrite and polyaniline powders were characterized using X-ray diffraction (XRD)
S Dianat et al.
Chemico-biological interactions, 215, 25-32 (2014-03-13)
The parent and nanosized starch, and lipid encapsulated K6[SiW11O39Co(H2O)]·nH2O (abbreviated as SEP, LEP and SiW11Co, respectively), as potent antitumor candidates, were synthesized and characterized by FT-IR spectroscopy, ICP, TG analysis, SEM and TEM images. The results show that the SiW11Co
Aya Watanabe et al.
Clinical science (London, England : 1979), 126(9), 671-678 (2013-11-22)
In the present study we sought to determine the effect of CoCl2, an inhibitor of PHD (prolyl hydroxylase domain protein), on the development of AAA (abdominal aortic aneurysm). AAA was induced in C57BL/6 mice by periaortic application of CaCl2 (AAA
John J Devlin et al.
Journal of medical toxicology : official journal of the American College of Medical Toxicology, 9(4), 405-415 (2013-11-14)
Safety concerns regarding cobalt-containing metal alloy hip prosthetics (Co-HP) have resulted in product recalls, a medical device alert, and issuance of guidance for clinicians. Recently, cases of suspected prosthetic hip-associated cobalt toxicity (PHACT) from Co-HP have been reported. Although little

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service