Merck
  • Clinical features, testing, and management of patients with suspected prosthetic hip-associated cobalt toxicity: a systematic review of cases.
Clinical features, testing, and management of patients with suspected prosthetic hip-associated cobalt toxicity: a systematic review of cases.
Journal of medical toxicology : official journal of the American College of Medical Toxicology (2013-11-14)
John J Devlin, Adam C Pomerleau, Jeffrey Brent, Brent W Morgan, Scott Deitchman, Michael Schwartz
ABSTRACT
Safety concerns regarding cobalt-containing metal alloy hip prosthetics (Co-HP) have resulted in product recalls, a medical device alert, and issuance of guidance for clinicians. Recently, cases of suspected prosthetic hip-associated cobalt toxicity (PHACT) from Co-HP have been reported. Although little is known about suspected PHACT, these patients may be referred to medical toxicologists for evaluation and management recommendations. We searched MEDLINE, EMBASE, and unpublished abstracts from toxicology scientific meetings for references relevant to PHACT. Authors independently screened publications for inclusion criteria: publication in English, human study population, subject(s) are symptomatic (except for isolated hip pain), and cobalt values in any matrix (blood, serum, urine, CSF, synovial fluid) available for review. Data from 10 cases are reviewed. Patients with suspected PHACT had findings consistent with cobalt toxicity, including thyroid, cardiac, and neurologic dysfunction. Signs and symptoms appeared between 3 and 72 months after arthroplasty (median 19 months). Neurologic symptoms were most common. Ancillary testing varied considerably. All patients had elevated cobalt levels in one or more matrices. Enhanced elimination was attempted in 27 % of patients. At this time, the information currently available regarding patients with suspected PHACT is inadequate to guide clinical decision making. No consensus has been reached regarding the management of Co-HP patients with systemic symptoms. Indications for chelation have not been established and require further study. Improved case definitions, improved surveillance, and controlled studies are needed to elucidate the scope of this problem and guide future investigations.
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Cobalt, foil, 25mm disks, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, 4mm disks, thickness 0.008mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.009mm, 99.9%
Cobalt, foil, 25mm disks, thickness 0.125mm, annealed, 99.9%
Cobalt, foil, 4mm disks, thickness 0.125mm, annealed, 99.9%
Cobalt, foil, 6mm disks, thickness 0.007mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.0125mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.006mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.008mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.015mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Cobalt, foil, 6mm disks, thickness 0.009mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.01mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.003mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.125mm, annealed, 99.9%
Cobalt, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, 8mm disks, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, 4mm disks, thickness 0.006mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.009mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.005mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.125mm, annealed, 99.9%