Skip to Content
Merck
All Photos(2)

Key Documents

GF59022592

Antimony

rod, 15mm, diameter 2.0mm, 99.99%

Synonym(s):

Antimony, SB007902

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sb
CAS Number:
Molecular Weight:
121.76
MDL number:
UNSPSC Code:
12141704
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99%

form

rod

manufacturer/tradename

Goodfellow 590-225-92

resistance

41.7 μΩ-cm

L × diam.

15 mm × 2.0 mm

bp

1635 °C (lit.)

mp

630 °C (lit.)

density

6.69 g/mL at 25 °C (lit.)

SMILES string

[Sb]

InChI

1S/Sb

InChI key

WATWJIUSRGPENY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Nash et al.
Journal of environmental monitoring : JEM, 2(2), 97-109 (2001-03-17)
Methodologies for the environmental analysis of total antimony and aqueous chemical speciation are critically reviewed, including preparation techniques for aqueous and solid matrices and the determination of solid state partitioning and recommendations are given for future research directions. Concentrations of
F Welle et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 28(1), 115-126 (2010-12-25)
Plastics bottles made from polyethylene terephthalate (PET) are increasingly used for soft drinks, mineral water, juices and beer. In this study a literature review is presented concerning antimony levels found both in PET materials as well as in foods and
Pankaj Sharma et al.
Acta pharmacologica Sinica, 29(8), 881-890 (2008-07-31)
Antimony, a natural element that has been used as a drug for over more than 100 years, has remarkable therapeutic efficacy in patients with acute promyelocytic leukemia. This review focuses on recent advances in developing antimony anticancer agents with an
A Léonard et al.
Mutation research, 366(1), 1-8 (1996-10-01)
The paper reviews the information available concerning the mutagenic, teratogenic and carcinogenic effects of antimony. A claim that antimony compounds could have mutagenic properties is based on insufficient and not particularly relevant data. Additional experiments, particularly with organic antimony compounds
Martin Wiener et al.
Journal of plastic, reconstructive & aesthetic surgery : JPRAS, 67(8), 1038-1044 (2014-06-15)
Recommended selective neck dissections for primary cutaneous tumours on the head and neck are based largely on clinical recurrence data acquired prior to the era of lymphatic mapping and sentinel lymph node (SLN) biopsy. This study aimed to examine lymphatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service