95372
Bismuth
granular, ≥99.99% trace metals basis
Synonym(s):
Bi
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
<0.1 mmHg ( 20 °C)
Assay
≥99.99% trace metals basis
form
granular
reaction suitability
core: bismuth
reagent type: catalyst
resistivity
129 μΩ-cm, 20°C
bp
1560 °C (lit.)
mp
271 °C (lit.)
272 °C
density
9.8 g/mL at 25 °C (lit.)
SMILES string
[Bi]
InChI
1S/Bi
InChI key
JCXGWMGPZLAOME-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Bismuth is a silvery, very brittle metal with a high metallic luster. In the periodic table, it is next to lead. Bismuth has a variety of applications, including medicine, materials synthesis, organic synthesis, and catalysis . It is used in the production of malleable iron and as a catalyst in the manufacture of acrylic fibers . Additionally, bismuth promotes noble metal-based catalysts for the selective oxidation of alcohols, aldehydes and carbohydrates with molecular oxygen in liquid phases.
Application
Bismuth granules are used in the preparation of bismuth nanoparticles by solution dispersion method. It is a precursor to synthesize bismuth-silicate glasses.
Bismuth is used as a precursor in the synthesis of Cu3BiS3 based solar device.
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Characterization of structural, thermal and mechanical properties of bismuth silicate glasses.
Bochentyn B, et al.
Journal of Non-Crystalline Solids, 439(2), 51-56 (2016)
Modification of carbon screen-printed electrodes by adsorption of chemically synthesized Bi nanoparticles for the voltammetric stripping detection of Zn (II), Cd (II) and Pb (II).
Rico MAG, et al.
Talanta, 80(2), 631-635 (2009)
The role of bismuth in the SOHIO process
Hanna TA
Coordination Chemistry Reviews, 248(5-6), 429-440 (2004)
The elements
Hammond CR
CRC Handbook of Chemistry and Physics, 81 (2000)
A simple way to prepare bismuth nanoparticles.
Zhao Y, et al.
Materials Letters, 58(5), 790-793 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service