Skip to Content
Merck
All Photos(2)

Key Documents

GF43011391

Lanthanum

foil, 100x100mm, thickness 0.125mm, as rolled, 99%

Synonym(s):

Lanthanum, LA000215

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
La
CAS Number:
Molecular Weight:
138.91
MDL number:
UNSPSC Code:
12141607
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

foil

manufacturer/tradename

Goodfellow 430-113-91

resistivity

54 μΩ-cm

bp

3464 °C (lit.)

mp

920 °C (lit.)

density

6.19 g/mL at 25 °C (lit.)

SMILES string

[La]

InChI

1S/La

InChI key

FZLIPJUXYLNCLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Declan de Freitas et al.
Seminars in dialysis, 20(4), 325-328 (2007-07-20)
Hyperphosphatemia is associated with increased morbidity and mortality in dialysis patients. Oral phosphate binders are necessary to control serum phosphate in patients eating a normal diet and undergoing peritoneal dialysis or thrice weekly hemodialysis. Until recently, none of the available
Stephen J P Damment et al.
Clinical pharmacokinetics, 47(9), 553-563 (2008-08-14)
Lanthanum carbonate is considered to be the most potent of a new generation of noncalcium phosphate binders used to treat hyperphosphataemia in chronic kidney disease (CKD), a condition associated with progressive bone and cardiovascular pathology and a markedly elevated risk
Amy Barton Pai et al.
Expert opinion on drug metabolism & toxicology, 5(1), 71-81 (2009-02-25)
Hyperphosphatemia is recognized as a principal mineral disorder in chronic kidney disease (CKD) that leads to the development of secondary hyperparathyroidism. Recent data indicate that hyperphosphatemia is associated with accelerated cardiac calcification and increased mortality in patients with CKD. Control
Mario Cozzolino et al.
Expert opinion on pharmacotherapy, 13(16), 2337-2353 (2012-10-05)
High phosphate levels are associated with unfavorable outcomes in ESRD. Recent data suggested that phosphate levels within the normal range are equally associated with poor outcomes in the community and CKD stage 3 - 4. Several concept papers support the
Marc E De Broe
Seminars in dialysis, 21(2), 142-144 (2008-02-06)
The recent recognition of gadolinium (Gd) administration as a cause of nephrogenic systemic fibrosis (NSF) is a growing concern. The similarity of Gd and lanthanum (La), based on their place in the table of Mendeljev, has led to assumptions that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service