Skip to Content
Merck
All Photos(2)

Key Documents

GF40338132

Boron

monofilament, 5m, diameter 0.2mm

Synonym(s):

Boron, B 005915

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B
CAS Number:
Molecular Weight:
10.81
MDL number:
UNSPSC Code:
12141907
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

foil

manufacturer/tradename

Goodfellow 403-381-32

resistivity

1.5E12 μΩ-cm, 20 °C

L × diam.

5 m × 0.2 mm

density

2.34 g/mL at 25 °C (lit.)

SMILES string

[B]

InChI

1S/B

InChI key

ZOXJGFHDIHLPTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ewan Galbraith et al.
Chemical Society reviews, 39(10), 3831-3842 (2010-09-08)
This critical review will explore some of the recent research into the use of boron based anion receptors and focus on those receptors that have been designed to transduce the binding event into a measurable output signal. In particular the
Kyoko Miwa et al.
Annals of botany, 105(7), 1103-1108 (2010-03-17)
The essentiality of boron (B) for plant growth was established > 85 years ago. In the last decade, it has been revealed that one of the physiological roles of B is cross-linking the pectic polysaccharide rhamnogalacturonan II in primary cell
Anthony R Martin et al.
Chemical Society reviews, 42(13), 5684-5713 (2013-03-30)
At the intersection of nucleic acid and boron chemistries lies a thriving world of possibilities. During the past decades, the merging of these research domains has led to fascinating discoveries in different fields ranging from material to medical sciences. In
Margaret Pallotta et al.
Nature, 514(7520), 88-91 (2014-07-22)
Environmental constraints severely restrict crop yields in most production environments, and expanding the use of variation will underpin future progress in breeding. In semi-arid environments boron toxicity constrains productivity, and genetic improvement is the only effective strategy for addressing the
Anne-Marie Alexander et al.
Chemical Society reviews, 39(11), 4388-4401 (2010-06-08)
Catalysts generated by the addition of carbon, nitrogen or phosphorus to transition metals have interesting properties and potential applications. The addition of carbon, nitrogen or phosphorus can lead to substantial modification of the catalytic efficacy of the parent metal and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service