Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Boron
Synonym(s):
Boron
Empirical Formula (Hill Notation):
B
CAS Number:
7440-42-8
Molecular Weight:
10.81
Products
Brand
Sigma-Aldrich (3)
Formula Weight
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Element
Quality Segment
Particle Size (µm)
Filter Products
Showing 1-20 of 33
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
15580
Boron,
≥95% (boron), amorphous powder
Expand
View Pricing
1.12070
Boron,
powder
Expand
View Pricing
266620
Boron,
crystalline, 1 cm, 99.7% trace metals basis
Expand
View Pricing
GF13726920
Boron,
powder, max. particle size 45 micron, weight 50 g, purity 98.%
Expand
View Pricing
GF72528239
Boron,
powder, 45 max. part. size (micron), weight 10 g, purity 98.%
Expand
View Pricing
GF95185222
Boron,
powder, mean particle size (fsss) 1 micron, weight 10 g, purity 92.1%
Expand
View Pricing
GF70012370
Boron,
powder, weight 100 g, mean particle size (fsss) 0.9 micron, purity 95+%
Expand
View Pricing
GF25620353
Boron,
granule, 15 mm nominal granule size, weight 50 g, purity 99%
Expand
View Pricing
GF03760095
Boron,
powder, mean particle size (fsss) 0.9 micron, weight 50 g, purity 95+%
Expand
View Pricing
GF53771610
Boron,
granule, 15 mm nominal granule size, weight 100 g, purity 99%
Expand
View Pricing
GF47837065
Boron,
powder, max. particle size 45 micron, weight 20 g, purity 98.%
Expand
View Pricing
GF96030086
Boron,
powder, mean particle size (fsss) 0.9 micron, weight 20 g, purity 95+%
Expand
View Pricing
GF55168241
Boron,
powder, mean particle size (fsss) 0.9 micron, weight 10 g, purity 95+%
Expand
View Pricing
GF28460752
Boron,
granule, 15 mm nominal granule size, weight 20 g, purity 99%
Expand
View Pricing
GF98152601
Boron,
powder, weight 20 g, mean particle size (fsss) 1 micron, purity 92.1%
Expand
View Pricing
GF51546292
Boron,
powder, mean particle size (fsss) 0.9 micron, weight 200 g, purity 95+%
Expand
View Pricing
GF40338132
Boron,
monofilament, 5m, diameter 0.2mm
Expand
View Pricing
GF34405297
Boron,
powder, mean particle size (fsss) 1 micron, weight 100 g, purity 92.1%
Expand
View Pricing
GF98835069
Boron,
monofilament, 5m, diameter 0.1mm
Expand
View Pricing
GF66774752
Boron,
monofilament, 0.080 mm diameter, length 50 m
Expand
View Pricing
Page 1 of 2
Page 1 of 2