Skip to Content
Merck
All Photos(1)

Key Documents

F3556

Sigma-Aldrich

Fluorinert FC-770

liquid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

Fluorinert FC-770,

form

liquid

Quality Level

color

colorless

viscosity

0.7-0.9 cSt(25 °C)

density

1.760-1.800 g/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C10F22/c11-1(12,3(15,16)5(19,20)7(23,24)9(27,28)29)2(13,14)4(17,18)6(21,22)8(25,26)10(30,31)32

InChI key

BPHQIXJDBIHMLT-UHFFFAOYSA-N

General description

Fluorinert belongs to the group of aliphatic compounds. It is considered as a pressure-transmitting medium because it is highly inert. Fluorinert Electronic Liquid FC-770 is a thermally stable, fully-fluorinated liquid that has ideal properties for use as a heat transfer fluid in a variety of industries. The inertness of Fluorinert FC-770 permits its use as a direct contact single and two-phase coolant in certain supercomputers and sensitive military electronics. Its high dielectric strength and electrical resistivity are ideal for applications in high voltage transformers and power electronics. In the semiconductor manufacturing industry, its wide liquid range makes Fluorinert liquid FC-770 suited for cooling ion implanters, dry etchers and CVD machines. Its low pour point also permits its use in thermal shock and test equipment.

Application

Fluorinert FC-770 has been used as an enema in a multimodality imaging study (MRC and endoscopy) for quantitative analysis of the effect of rapamycin on colonic adenomas in mouse model. In the same context it has also been used for anatomic and functional (dynamic contrast-enhanced (DCE)- MRI) assessment of colonic polyps in multiple intestinal neoplasia mice. It may also be used a compound to calibrate electron ionization (EI) mass spectroscopy.

Packaging

Packaged and distributed by Sigma-Aldrich

Other Notes

Fluorinert FC-770 is a thermally stable, fully-fluorinated liquid with high dielectric strength and electrical resistivity.

Legal Information

Fluorinert is a trademark of 3M Company

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorinert as a pressure-transmitting medium for high-pressure diffraction studies.
Varga T, et al.
The Review of Scientific Instruments, 74(10), 4564-4566 (2003)
In vivo MRI virtual colonography in a mouse model of colon cancer.
Ileva L V, et al.
Nature Protocols, 9(11), 2682-2682 (2014)
Reactions of perfluorotri-n-butylamine fragment ions in the quadrupole ion trap: the origin of artefacts in the perfluorotri-n-butylamine calibration spectrum.
Creaser C S, et al.
Rapid Communications in Mass Spectrometry, 14(6), 538-540 (2000)
Dogukan Mizrak et al.
Cell reports, 31(12), 107805-107805 (2020-06-25)
In the adult ventricular-subventricular zone (V-SVZ), neural stem cells (NSCs) generate new olfactory bulb (OB) neurons and glia throughout life. To map adult neuronal lineage progression, we profiled >56,000 V-SVZ and OB cells by single-cell RNA sequencing (scRNA-seq). Our analyses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service