Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
CAS Number:
NACRES:
NA.28
PubChem Substance ID:
UNSPSC Code:
15121530
EC Number:
206-421-8
MDL number:
description
used in density gradient studies
density
1.93 g/mL at 25 °C (lit.)
SMILES string
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)N(C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
InChI
1S/C15F33N/c16-1(17,4(22,23)10(34,35)36)7(28,29)13(43,44)49(14(45,46)8(30,31)2(18,19)5(24,25)11(37,38)39)15(47,48)9(32,33)3(20,21)6(26,27)12(40,41)42
InChI key
AQZYBQIAUSKCCS-UHFFFAOYSA-N
Application
For use as a high density chase solution in density gradient studies.
Fluorinert™ FC-70 has been used to reduce the adherence of large trypsin:N-amidinopiperidine crystals to the well. It has also been used as a fluorocarbon oil for microinjection of oocytes.
Packaging
Packaged and distributed by Sigma-Aldrich
Other Notes
Fluorinert FC-70 is a liquid mixture of completely fluorinated aliphatic compounds.
Legal Information
Fluorinert is a trademark of 3M Company
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Quantitative microinjection of oocytes, eggs, and embryos
Jaffe LA and Terasaki M
Methods in Cell Biology, 74, 219-242 (2004)
Alexandra Colin et al.
The Journal of cell biology, 219(3) (2020-01-18)
Nucleus centering in mouse oocytes results from a gradient of actin-positive vesicle activity and is essential for developmental success. Here, we analyze 3D model simulations to demonstrate how a gradient in the persistence of actin-positive vesicles can center objects of
Intriguing role of water in protein-ligand binding studied by neutron crystallography on trypsin complexes
Schiebel J, et al.
Nature Communications, 9 (2018)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| F9880-100ML | 04061833622131 |
| F9880-25ML | 04061837840739 |