Skip to Content
Merck
All Photos(1)

Key Documents

DWK986734

WHEATON® liquid scintillation vial with attached unlined PS cap

lips on vial transparent PET bottle, capacity (20 mL), screw cap

Synonym(s):

986734

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.

material

lips on vial transparent PET bottle
white polyethylene cap

feature

with cap

packaging

case of 500 ea Trays of 100 vials are shrink-wrapped in plastic for protection against contamination.

manufacturer/tradename

WHEATON® 986734

bottle diam. × H × W

(27 mm) (61 mm) (302 mm)

capacity

(20 mL)

fitting

22-400 neck
22-400

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service