DWK292432859
Bottle Tag, For Gl 45 Duran® Media Bottles Bottle Tag
blue silicone bottle
Synonym(s):
292432859
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
Recommended Products
material
blue silicone bottle
packaging
pkg of 20 ea
manufacturer/tradename
DURAN® 292432859
Looking for similar products? Visit Product Comparison Guide
General description
The colourful GL 45 Bottle Tags offer an elegant solution to the marking of laboratory bottles. The Bottle Tags can be easily and securely attached around the neck of the YOUTILITY bottle, even with the screw cap in place. The Bottle Tags feature a graspable tab that makes them easy to remove with bare or gloved hands.
Application
easy customisation and unambiguous bottle identification.
Features and Benefits
for GL 45 bottles
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service