Skip to Content
Merck
All Photos(3)

Key Documents

D5146

Sigma-Aldrich

Diazolidinyl urea

≥95%

Synonym(s):

1-[1,3-Bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1,3-bis(hydroxymethyl)urea, Diazolidinylurea, N-Hydroxymethyl-N-[1,3-di(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-N′-hydroxymethylurea, N-[1,3-Bis(hydroxymethyl)-2,5-dioxo-4-imidazolidinyl]-N,N′-bis(hydroxymethyl)urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N4O7
CAS Number:
Molecular Weight:
278.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

storage temp.

2-8°C

SMILES string

OCNC(=O)N(CO)C1N(CO)C(=O)N(CO)C1=O

InChI

1S/C8H14N4O7/c13-1-9-7(18)10(2-14)5-6(17)12(4-16)8(19)11(5)3-15/h5,13-16H,1-4H2,(H,9,18)

InChI key

SOROIESOUPGGFO-UHFFFAOYSA-N

General description

Diazolidinyl urea is urea derivative that is commonly used as a preservative and antimicrobial agent in cosmetics and personal care products.

Application

  • Impact of cosmetic preservatives on skin cells: A study examined the effects of various cosmetic preservatives, including Diazolidinyl Urea, on healthy human skin cells, highlighting the cellular responses and potential implications for skin health (Głaz et al., 2023).
  • Formaldehyde releasers in personal care: An extensive clinical review discussed allergic contact dermatitis caused by formaldehyde and formaldehyde releasers such as Diazolidinyl Urea, providing insights into allergic reactions and safety concerns in dermatological practice (Goossens and Aerts, 2022).
  • Carcinogenic potential of cosmetic ingredients: Research on potential carcinogens in makeup cosmetics included Diazolidinyl Urea, emphasizing the safety and regulatory aspects concerning its use in cosmetic formulations (Balwierz et al., 2023).
  • Patch testing for allergen sensitivity: A study integrated Diazolidinyl Urea in a patch test series to evaluate skin sensitization and allergic reactions in patients, aiding in the assessment of its allergenic potential (Bizjak et al., 2022).

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S D Winslow et al.
Environmental science & technology, 35(20), 4103-4110 (2001-11-01)
Preservation of chemical analytes in drinking water samples is necessary to obtain accurate information concerning contaminant occurrence. Sample preservation to prevent biodegradation is important for most samples and analytes. With the unique demands of environmental methods, it is not always
A T Karlberg et al.
Contact dermatitis, 38(1), 20-28 (1998-03-21)
Reliable and uncomplicated methods for detection of free formaldehyde in products preserved with formaldehyde donors are desirable to decrease the risk of allergic contact dermatitis. The aim of this study was to develop a method that could be used in
Diazolidinyl urea: incidence of sensitivity, patterns of cross?reactivity and clinical relevance.
Hectorne KJ & Fransway AF
Contact Dermatitis, 30(1), 16-19 (1994)
Daeryl E Amponin et al.
BMC ophthalmology, 20(1), 28-28 (2020-01-17)
Corneal infections with antibiotic-resistant microorganisms are an increasingly difficult management challenge and chemically or photochemically cross-linking the cornea for therapy presents a unique approach to managing such infections since both direct microbial pathogens killing and matrix stabilization can occur simultaneously.
Claus Zachariae et al.
Contact dermatitis, 53(5), 268-277 (2005-11-15)
The elicitation potential of the cosmetic preservative diazolidinyl urea was studied in formaldehyde- and diazolidinyl urea-sensitized volunteer patients using a stepwise controlled exposure design. The test product was a facial moisturizer, preserved with varying concentrations of diazolidinyl urea, ranging from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service