46885-U
ProClin™ 950
pkg of 5 mL, ProClin™ preservative for use in IVD manufacturing, MIT 9.5 %
Synonym(s):
9.5% solution of 2-Methyl-4-isothiazolin-3-one, 2-Methyl-4-isothiazolin-3-one solution
About This Item
Recommended Products
Quality Level
form
liquid
shelf life
3 yr
composition
MIT, 9.5%
packaging
pkg of 5 mL
parameter
<90 °C operating temperature
color
colorless to light yellow
matrix
Water
operating pH range
2-12
pH range
3-6
bp
100 °C
mp
-20--15 °C
solubility
water: soluble at 20 °C
density
1.020 g/cm3 at 25 °C
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service