Skip to Content
Merck
All Photos(1)

Documents

D183601

Sigma-Aldrich

2,5-Dimethylpyrrole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N
CAS Number:
Molecular Weight:
95.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.505 (lit.)

bp

165 °C/740 mmHg (lit.)

density

0.935 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)[nH]1

InChI

1S/C6H9N/c1-5-3-4-6(2)7-5/h3-4,7H,1-2H3

InChI key

PAPNRQCYSFBWDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongyin Yin et al.
PloS one, 8(9), e76011-e76011 (2013-10-08)
The formation of pyrrole adducts might be responsible for peripheral nerve injury caused by n-hexane. The internal dose of pyrrole adducts would supply more information for the neurotoxicity of n-hexane. The current study was designed to investigate the tissue distributions
Ireneusz Nowak et al.
Organic letters, 5(18), 3345-3348 (2003-08-29)
[reaction: see text] Protection of the amino group of adenine and guanine nucleosides was effected by heating the substrates in 2,5-hexanedione. The resulting 2,5-dimethylpyrrole adducts were stable toward bases but were hydrolyzed with TFA/H(2)O to regenerate the amino function.
Xianjie Li et al.
PloS one, 13(12), e0209939-e0209939 (2019-01-01)
Pyrrole adducts are specific reaction products of 2,5-hexadione (2,5-HD) in vivo and are considered highly relevant to the pathogenesis of peripheral nerve impairments after exposure to n-hexane, though the exact mechanism remains unclear. In this study, 40 male Wistar rats
Xianjie Li et al.
Neurotoxicology, 78, 11-20 (2020-02-12)
n-Hexane has been reported to induce serious peripheral neuropathy in workers. Pyrrole adducts are the unique reaction products of n-hexane in organisms and have been demonstrated to be critical to n-hexane neuropathy. Our previous studies have demonstrated that pyrrole adducts
Yoshiteru Matsumoto et al.
Physical chemistry chemical physics : PCCP, 13(31), 13962-13971 (2011-06-16)
N-H···π hydrogen-bonded (H-bonded) structures were studied by applying vibrational spectroscopy to self-aggregate clusters of 2,5-dimethylpyrrole (DMPy) and its binary clusters with pyrrole (Py). The NH stretching vibrations of jet-cooled clusters were observed by IR cavity ringdown spectroscopy. A combination of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service