Skip to Content
Merck
All Photos(3)

Documents

131709

Sigma-Aldrich

Pyrrole

reagent grade, 98%

Synonym(s):

Azole, Divinylenimine, Imidole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N
CAS Number:
Molecular Weight:
67.09
Beilstein:
1159
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39151603
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Quality Level

vapor density

2.31 (vs air)

Assay

98%

form

liquid

refractive index

n20/D 1.508 (lit.)

bp

131 °C (lit.)

mp

−23 °C (lit.)

density

0.967 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

c1cc[nH]c1

InChI

1S/C4H5N/c1-2-4-5-3-1/h1-5H

InChI key

KAESVJOAVNADME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyrrole is a heterocyclic aromatic compound that can undergo electrophilic aromatic substitution.

Application

Pyrrole was used in the electropolymerisation of macroporous conducting polymer films. It was also used to study the hydrogen-bond mediated coupling of 1,2,3-triazole to pyrrole.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guillaume Lautrette et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 20(6), 1547-1553 (2014-01-10)
Starting from a previously described aromatic oligoamide helically folded capsule that binds tartaric acid with high affinity and diastereoselectivity, we demonstrate the feasibility of the direct in situ modification of the helix backbone, which results in a conformational change that
Jian Wen et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 20(4), 974-978 (2014-01-01)
Hydrogen-bond mediated coupling of 1,2,3-triazoles to indoles and pyrroles results in N2 selective functionalization of the triazole moiety in moderate to excellent yields. The reaction was tolerant of un-, mono- and disubstituted triazoles and was applied to synthesize tryptophan derived
Electrochemical syntheses of highly ordered macroporous conducting polymers grown around self-assembled colloidal templates.
Bartlett PN, et al.
Journal of Materials Chemistry, 11(3), 849-853 (2001)
Pyrrole
Chen MZ and Tsai AS
eEROS (Encyclopedia of Reagents for Organic Synthesis), 1-4 (2001)
Sheng He et al.
Biomaterials, 258, 120285-120285 (2020-08-12)
Myocardial fibrosis, resulting from ischemic injury, increases tissue resistivity in the infarct area, which impedes heart synchronous electrical propagation. The uneven conduction between myocardium and fibrotic tissue leads to dys-synchronous contraction, which progresses towards ventricular dysfunction. We synthesized a conductive

Articles

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service