Skip to Content
Merck
All Photos(1)

Documents

D151203

Sigma-Aldrich

4,4′-Dimethylbiphenyl

97%

Synonym(s):

p,p′-Bitoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4C6H4CH3
CAS Number:
Molecular Weight:
182.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

bp

295 °C (lit.)

mp

118-120 °C (lit.)

SMILES string

Cc1ccc(cc1)-c2ccc(C)cc2

InChI

1S/C14H14/c1-11-3-7-13(8-4-11)14-9-5-12(2)6-10-14/h3-10H,1-2H3

InChI key

RZTDESRVPFKCBH-UHFFFAOYSA-N

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyou Suga et al.
The journal of physical chemistry. A, 109(44), 10168-10175 (2006-07-15)
Photooxygenation of 4,4'-dimethybiphenyl with oxygen occurs efficiently in the presence of 9-phenyl-10-methylacridinium perchlorate (AcrPh(+)ClO(4)(-)) under visible light irradiation in O(2)-saturated chloroform (CHCl(3)) to yield 4-(4'-methylphenyl)benzaldehyde as a main oxygenated product. Prolonged photoirradiation afforded the further oxygenated product, 4,4'-diformylbiphenyl. The reactive
K R Iyer et al.
Biochemistry, 36(23), 7136-7143 (1997-06-10)
Intramolecular isotope effects associated with the benzylic hydroxylation of a series of selectively deuterated isomeric xylenes and 4,4'-dimethylbiphenyl as catalyzed by various rat liver microsomal preparations and CYP2B1 were determined. Substrate analogs in which each methyl group contained either one
K R Henne et al.
Biochemistry, 40(29), 8597-8605 (2001-07-18)
The active site topography of rabbit CYP4B1 has been studied relative to CYP2B1 and CYP102 using a variety of aromatic probe substrates. Oxidation of the prochiral substrate cumene by CYP4B1, but not CYP2B1 or CYP102, resulted in the formation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service