Skip to Content
Merck

230950

Ciglitazone

A potent thiazolinedione (TDZ) type anti-hyperglycemic agent and a selective PPARγ agonist (EC50 = 3 µM).

Synonym(s):

Ciglitazone, (±)-5-[4-(1-Methylcyclohexylmethoxy)benzyl]thiazolidine-2,4-dione, PPAR Agonist II, PPARγ Agonist I

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H23NO3S
CAS Number:
Molecular Weight:
333.45
NACRES:
NA.77
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Quality level:
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

description

RTECS - XJ5813700

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

tan

solubility

DMSO: ≥25 mg/mL, ethanol: ≥25 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C18H23NO3S/c1-18(9-3-2-4-10-18)12-22-14-7-5-13(6-8-14)11-15-16(20)19-17(21)23-15/h5-8,15H,2-4,9-12H2,1H3,(H,19,20,21)

InChI key

YZFWTZACSRHJQD-UHFFFAOYSA-N

General description

A potent thiazolinedione (TDZ) type anti-hyperglycemic agent and a selective peroxisome proliferator-activated receptor γ (PPARγ) agonist (EC50 = 3 µM).

Biochem/physiol Actions

Cell permeable: no
EC50 = 3 µM as PPARγ agonist
Primary Target
Selective PPARγ agonist
Product does not compete with ATP.
Reversible: no

Preparation Note

Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Jha, R.J. 1999. Clin. Exp. Hypertens. 21, 157.
Xin, X., et al. 1999. J. Biol. Chem. 274, 9116.
Lohrke, B., et al. 1998. J. Endocrinol. 159, 429.
Willson, T.M., et al. 1996. J. Med. Chem. 39, 665.
Cantello, B.C., et al. 1994. J. Med. Chem. 37, 3977.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Yingyi Zhang et al.
Cell reports, 36(5), 109481-109481 (2021-08-05)
Preadipocytes dynamically produce sensory cilia. However, the role of primary cilia in preadipocyte differentiation and adipose homeostasis remains poorly understood. We previously identified transition fiber component FBF1 as an essential player in controlling selective cilia import. Here, we establish Fbf1tm1a/tm1a



Global Trade Item Number

SKUGTIN
230950-5MG04055977218312