Skip to Content
Merck
All Photos(2)

Documents

C115002

Sigma-Aldrich

Cyclopentylamine

99%

Synonym(s):

Aminocyclopentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9NH2
CAS Number:
Molecular Weight:
85.15
Beilstein:
635706
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.450 (lit.)

bp

106-108 °C (lit.)

density

0.863 g/mL at 25 °C (lit.)

SMILES string

NC1CCCC1

InChI

1S/C5H11N/c6-5-3-1-2-4-5/h5H,1-4,6H2

InChI key

NISGSNTVMOOSJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cycloheptylamineis a versatile compound that has several useful applications in organicsynthesis. Its ability to act as a building block, catalyst, and reagent makesit a valuable tool for the development of new organic compounds. It is alsoused as a reagent for the functionalization of organic molecules.

Application

Cycloheptylamine is a building block for the formation of an ABX3-typed perovskite structure compound [C5H9–NH3][CdCl3].

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

52.7 °F - closed cup

Flash Point(C)

11.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Room-temperature ABX 3-typed molecular ferroelectric:[C 5 H 9-NH 3][CdCl 3]
Zhang, Yi and Ye, et al.
Inorganic chemistry frontiers, 1, 118-123 (2014)
M S Sansom et al.
Protein engineering, 6(1), 65-74 (1993-01-01)
The influenza A M2 protein forms cation-selective ion channels which are blocked by the anti-influenza drug amantadine. A molecular model of the M2 channel is presented in which a bundle of four parallel M2 transbilayer helices surrounds a central ion-permeable
Maris Vilums et al.
Journal of medicinal chemistry, 56(19), 7706-7714 (2013-09-14)
Preclinical models of inflammatory diseases (e.g., neuropathic pain, rheumatoid arthritis, and multiple sclerosis) have pointed to a critical role of the chemokine receptor 2 (CCR2) and chemokine ligand 2 (CCL2). However, one of the biggest problems of high-affinity inhibitors of
D G Craciunescu et al.
In vivo (Athens, Greece), 1(4), 229-234 (1987-07-01)
Ten new Pt (II) complexes were synthesized and tested as potential antitumor drugs in vitro on KB human tumour cell line, and in vivo against four experimental tumour systems (P388, L1210, ADJ/PC6A and Yoshida sarcoma). The complexes contained two primary
M J Comin et al.
Nucleosides & nucleotides, 18(10), 2219-2231 (2000-01-05)
Purine carbanucleosides built on a 6-oxabicyclo[3.1.0]hexane template were synthesized from readily available 2-cyclopentenone employing a Mitsunobu reaction to incorporate the base onto the carbocyclic ring. Both adenosine and guanosine analogues exhibited moderate antiviral activity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service