Key Documents
BR721305
BRAND® counting chamber BLAUBRAND® Nageotte pattern
without clips, double ruled
Synonym(s):
BLAUBRAND®, Cell culture, Counting Chambers, Nageotte
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
material
colorless
glass
sterility
non-sterile
feature
without clips
manufacturer/tradename
BRAND 721305
wells
1
General description
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.The square area of 100 mm² is subdivided into 40 rectangles with an area of 2.5 mm² each.This chamber is commonly used for counting cells in the cerebro-spinal fluid, or for counting nematodes.
Features and Benefits
• BLAUBRAND® quality• Outstanding measurement precision• 100% certified quality
Packaging
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)