Key Documents
BR717810
BRAND® counting chamber BLAUBRAND® Neubauer improved
without clips, bright-line
Synonym(s):
BLAUBRAND®, Cell culture, Counting Chambers, Neubauer, ldpe bottle, polyethylene bottles, polyethylene containers
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
material
colorless
glass
sterility
non-sterile
feature
without clips
manufacturer/tradename
BRAND 717810
L × W × H
24 mm × 26 mm × 0.4 mm
chamber size
0.1 mm , depth
wells
1
General description
include 2 hemocytometer cover glasses. The dimensions are 24mm L x 26mm W x 0.4mm H. They are also sold separately as item BR723014.
Features and Benefits
  • BLAUBRAND® counting chambers are precision measuring instruments, used to determine the number of particles per volume unit of a suspension.
  • Same ruling as improved Neubauer, but with rhodium coated chamber bottom.
  • Rulings are engraved into the rhodium layer and appear bright under normal microscope settings.
  • By altering the contrast, the microscope image can be reversed, so that the rulings appear brighter or darker, as required.
Packaging
Include 2 hemocytometer cover glasses. The dimensions are 24mm L x 26mm W x 0.4mm H. They are also sold separately as item BR723014.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Jordan Gulli et al.
Biotechnology and bioengineering, 117(6), 1696-1709 (2020-02-27)
A barrier to cost-efficient biomanufacturing is the instability of engineered genetic elements, such as plasmids. Instability can also manifest at the whole-genome level, when fungal dikaryons revert to parental species due to nuclear segregation during cell division. Here, we show
Raffaele Dotolo et al.
Journal of cellular physiology, 231(3), 551-557 (2015-09-04)
Prolyl endopeptidase (PREP) is a serine protease which has been implicated in many biological processes, such as the maturation and degradation of peptide hormones and neuropeptides, learning and memory, cell proliferation and differentiation, and glucose metabolism. A small number of