Skip to Content
Merck
All Photos(1)

Documents

414883

Sigma-Aldrich

Hexyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5NCO
CAS Number:
Molecular Weight:
127.18
Beilstein:
1751836
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.421 (lit.)

bp

162-164 °C (lit.)

density

0.873 g/mL at 25 °C (lit.)

SMILES string

CCCCCCN=C=O

InChI

1S/C7H13NO/c1-2-3-4-5-6-8-7-9/h2-6H2,1H3

InChI key

ANJPRQPHZGHVQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexyl isocyanate has been investigated for its antigenicity in the guinea pig animal model of hapten-specific respiratory hypersensitivity. Polymerization of n-hexyl isocyanate using rare earth tris(2,6-di-tert-butyl-4-methylphenolate) [Ln(OAr)3] as initiator has been studied.

Application

Hexyl isocyanate may be employed for the synthesis of following:
  • rigid-rod, helical isocyanate-based macromonomers
  • rod-coil-rod triblock copolymers with 2-vinylpyridine, by living anionic polymerization
  • chiral poly(n-hexyl isocyanate) (PHIC) macromonomers, by living anionic polymerization

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis and Self-Assembly Studies of Amphiphilic Poly (n-hexyl isocyanate)-b lock-poly (2-vinylpyridine)-b lock-poly (n-hexyl isocyanate) Rod-Coil-Rod Triblock Copolymer.
Rahman M, et al.
Macromolecules, 39(15), 5009-5014 (2006)
Boer Liu et al.
Molecules (Basel, Switzerland), 26(15) (2021-08-08)
This work reveals the influence of pendant hydrogen bonding strength and distribution on self-assembly and the resulting thermomechanical properties of A-AB-A triblock copolymers. Reversible addition-fragmentation chain transfer polymerization afforded a library of A-AB-A acrylic triblock copolymers, wherein the A unit
Pulmonary hypersensitivity to hexyl isocyanate-ovalbumin aerosol in guinea pigs.
M H Karol et al.
Toxicology and applied pharmacology, 51(1), 73-80 (1979-10-01)
Chiroptical Properties of Graft Copolymers Containing Chiral Poly (n-hexyl isocyanate) as a Side Chain.
Shah PN, et al.
Macromolecules, 44(20), 7917-7925 (2011)
Garret M Miyake et al.
Journal of the American Chemical Society, 134(34), 14249-14254 (2012-08-16)
The synthesis of rigid-rod, helical isocyanate-based macromonomers was achieved through the polymerization of hexyl isocyanate and 4-phenylbutyl isocyanate, initiated by an exo-norbornene functionalized half-titanocene complex. Sequential ruthenium-mediated ring-opening metathesis polymerization of these macromonomers readily afforded well-defined brush block copolymers, with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service