Skip to Content
Merck
All Photos(1)

Documents

B92303

Sigma-Aldrich

4-tert-Butylcyclohexanone

99%

Synonym(s):

4-(1,1-Dimethylethyl)cyclohexanone, 4-tert-Butylcyclohexanone, p-tert-Butylcyclohexanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H9(=O)
CAS Number:
Molecular Weight:
154.25
Beilstein:
507309
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

crystals

bp

113-116 °C/20 mmHg (lit.)

mp

47-50 °C (lit.)

SMILES string

CC(C)(C)C1CCC(=O)CC1

InChI

1S/C10H18O/c1-10(2,3)8-4-6-9(11)7-5-8/h8H,4-7H2,1-3H3

InChI key

YKFKEYKJGVSEIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Zhenlan Fang et al.
Angewandte Chemie (International ed. in English), 54(25), 7234-7254 (2015-06-04)
Defect engineering in metal-organic frameworks (MOFs) is an exciting concept for tailoring material properties, which opens up novel opportunities not only in sorption and catalysis, but also in controlling more challenging physical characteristics such as band gap as well as
Andrew F M Johnstone et al.
Toxicology in vitro : an international journal published in association with BIBRA, 69, 104989-104989 (2020-09-04)
The Hard-Soft Acid and Base hypothesis can be used to predict the potential bio-reactivity (electrophilicity) of a chemical with intracellular proteins, resulting in neurotoxicity. Twelve chemicals predicted to be neurotoxic were evaluated in vitro in rat dorsal root ganglia (DRG)
Selvedin Telalović et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(7), 2077-2088 (2011-01-25)
Bimetallic three-dimensional amorphous mesoporous materials, Al-Zr-TUD-1 materials, were synthesised by using a surfactant-free, one-pot procedure employing triethanolamine (TEA) as a complexing reagent. The amount of aluminium and zirconium was varied in order to study the effect of these metals on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service