Skip to Content
Merck
All Photos(1)

Key Documents

B8154

Sigma-Aldrich

Baccatin III

≥95% (HPLC)

Synonym(s):

O-De[(2R,3S)-3-(Benzoylamino)-2-hydroxy-3-phenylpropanoyl]paclitaxel

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H38O11
CAS Number:
Molecular Weight:
586.63
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥95% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

CC(=O)O[C@H]1C(=O)[C@]2(C)[C@@H](O)C[C@H]3OC[C@@]3(OC(C)=O)[C@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](O)C(C)=C1C5(C)C

InChI

1S/C31H38O11/c1-15-19(34)13-31(38)26(41-27(37)18-10-8-7-9-11-18)24-29(6,20(35)12-21-30(24,14-39-21)42-17(3)33)25(36)23(40-16(2)32)22(15)28(31,4)5/h7-11,19-21,23-24,26,34-35,38H,12-14H2,1-6H3/t19-,20-,21+,23+,24-,26-,29+,30-,31+/m0/s1

InChI key

OVMSOCFBDVBLFW-VHLOTGQHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to paclitaxel

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Da Cheng Hao et al.
PloS one, 6(6), e21220-e21220 (2011-07-07)
Illumina second generation sequencing is now an efficient route for generating enormous sequence collections that represent expressed genes and quantitate expression level. Taxus is a world-wide endangered gymnosperm genus and forms an important anti-cancer medicinal resource, but the large and
Elizabeth M Heider et al.
Physical chemistry chemical physics : PCCP, 9(46), 6083-6097 (2008-01-03)
The three-dimensional structure of a unique polymorph of the anticancer drug paclitaxel (Taxol) is established using solid state NMR (SSNMR) tensor ((13)C & (15)N) and heteronuclear correlation ((1)H-(13)C) data. The polymorph has two molecules per asymmetric unit (Z' = 2)
Hiroshi Izumi et al.
The Journal of organic chemistry, 73(6), 2367-2372 (2008-02-16)
The comparison between measured and conformer-weighted calculated VCD spectra of the baccatin III ring of paclitaxel and visualization of the conformations using the new code for structure-activity relationships are reported for the first time. The VCD spectrum of paclitaxel closely
Mark E Ondari et al.
The Journal of organic chemistry, 74(5), 2186-2188 (2009-02-10)
A one-pot trisilylation step to protect three hydroxyl groups of baccatin III (1), followed by hydride ester cleavage and base hydrolysis of a triethylsilyl ether at C13, provides efficient access to a key intermediate 9 (top path). This route removes
Takayuki Doi et al.
Chemistry, an Asian journal, 1(3), 370-383 (2007-04-19)
A 36-step synthesis was carried out in automated synthesizers to provide a synthetic key intermediate of taxol. A key step involved a microwave-assisted alkylation reaction to construct the ABC ring system from an AC precursor. Subsequent formation of the D

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service