Skip to Content
Merck
All Photos(2)

Key Documents

432725

Sigma-Aldrich

2,4-Dimethoxybenzylamine

98%

Synonym(s):

(2,4-Dimethoxyphenyl)methanamine, 1-(2,4-Dimethoxyphenyl)methanamine, 2,4-Dimethoxybenzenemethanamine, 2,4-Dimethyloxybenzylamine, [(2,4-Dimethoxyphenyl)methyl]amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2NH2
CAS Number:
Molecular Weight:
167.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.549 (lit.)

bp

140 °C/1 mmHg (lit.)

density

1.113 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

COc1ccc(CN)c(OC)c1

InChI

1S/C9H13NO2/c1-11-8-4-3-7(6-10)9(5-8)12-2/h3-5H,6,10H2,1-2H3

InChI key

QOWBXWFYRXSBAS-UHFFFAOYSA-N

General description

2,4-Dimethoxybenzylamine can be preprepared by reduction (NaBH4 , BF3.OEt2, THF) of 2,4-dimethoxybenzonitrile.

Application

2,4-Dimethoxybenzylamine is an amine nucleophile used to investigate the 1,4- reactivity of 5-bromo-2-indene-1-one. It may be used in the following studies:
  • As an ammonia equivalent in the concise synthesis of a series of 2,4,5-trisubstituted oxazoles, via a tandem Ugi/Robinson-Gabriel reaction sequence.
  • Total synthesis of (-)-muraymycin (MRY) D2 and its epimer, the antibacterial nucleoside natural product.
  • Two-step synthesis of amide derivatives of uracil polyoxin C (UPOC) methyl ester using the Ugi reaction.
  • Synthesis of N-hydroxythiourea.
  • Synthesis of anti-HIV-1 agents.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shinichi Imamura et al.
Chemical & pharmaceutical bulletin, 52(1), 63-73 (2004-01-08)
A novel lead compound, N-(3-[4-(4-fluorobenzoyl)piperidin-1-yl]propyl)-1-methyl-5-oxo-N-phenylpyrrolidine-3-carboxamide (1), was identified as a CCR5 antagonist by high-throughput screening using [(125)I]RANTES and CCR5-expressing CHO cells. The IC(50) value of 1 was 1.9 microM. In an effort to improve the binding affinity of 1, a
Keith J Stanger et al.
Journal of combinatorial chemistry, 8(3), 435-439 (2006-05-09)
We describe parallel/combinatorial, solid-phase, supported synthesis of diverse hydroxamates using a common intermediate, an N-derivatized, O-linked hydroxylamine. The method allows the concurrent synthesis of both N-alkyl and N-H hydroxamates and is compatible with a wide range of chemical transformations. The
M Sato et al.
Journal of medicinal chemistry, 19(2), 336-337 (1976-02-01)
The synthesis of the title compound (1) was accomplished by the conversion of 2,4-dimethoxybenzylamine (2) into an isothiocyanate (3) using thiocarbonyl diimidazole. Treatment of 3 with hydroxylamine and removal of the DMB group with trifluoroacetic acid gave 1. N-Hydroxythiourea (1)
Arthur Y Shaw et al.
Tetrahedron letters, 53(15), 1998-2000 (2013-04-06)
This Letter discloses a novel concise synthesis of a series of 2,4,5-trisubstituted oxazoles via a tandem Ugi/Robinson-Gabriel sequence. Herein, 2,4-dimethoxybenzylamine
Andrew Plant et al.
The Journal of organic chemistry, 74(13), 4870-4873 (2009-05-22)
A simple, two-step synthesis of amide derivatives of uracil polyoxin C (UPOC) methyl ester using the Ugi reaction is described. The four components employed in the Ugi reaction are 2',3'-isopropylidine-protected uridine-5'-aldehyde, 2,4-dimethoxybenzylamine, an isoxazolecarboxylic acid, and the convertible isonitrile N-(2-{[(tert-butyldimethylsilyl)oxy]methyl}phenyl)carbonitrile.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service