Skip to Content
Merck
All Photos(2)

Key Documents

932116

Sigma-Aldrich

9-(Naphthalen-1-yl)-10-(naphthalen-2-yl)anthracene

≥99% (HPLC)

Synonym(s):

Α,Β-ADN, 10-(1-Naphthalenyl)-9-(2-naphthalenyl)anthracene, 3,3′′,5,5′′-Tetra(3-pyridyl)-1,1′:3′,1′′-terbenzene, 9-(1-Naphthyl)-10-(2-naphthyl) anthracene, 9-(1-Naphthyl)-10-(2-naphthyl)anthracene, 9-(1-naphthyl)-10-(2-naphthyl) anthracene, 9-(Naphthalen-1-yl)-10-(naphthalen-2-yl)anthracene, 9-Naphthalen-1-yl-10-naphthalen-2-yl-anthracene, 9-Naphthalen-1-yl-10-naphthalen-2-ylanthracene, a,beta-AND

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H22
CAS Number:
Molecular Weight:
430.54
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.23
Assay:
≥99% (HPLC)
grade:
sublimed grade
solubility:
chloroform: soluble
dichloromethane: soluble

grade

sublimed grade

Quality Level

Assay

≥99% (HPLC)

loss

0.5%, .> 250 °C (weight loss)

solubility

chloroform: soluble
dichloromethane: soluble

SMILES string

c21c(cccc2c3c4c(c(c7c3cccc7)c5cc6c(cc5)cccc6)cccc4)cccc1

InChI

1S/C34H22/c1-2-12-25-22-26(21-20-23(25)10-1)33-29-15-5-7-17-31(29)34(32-18-8-6-16-30(32)33)28-19-9-13-24-11-3-4-14-27(24)28/h1-22H

Application

9-(Naphthalen-1-yl)-10-(naphthalen-2-yl)anthracene can be used as a host material in the emissive layer of organic light emitting diodes (OLEDs) devices. It can also be used as a phosphorescent emitter in OLEDs when doped with appropriate metals such as iridium or platinum.
It has been shown to have good stability and exhibits a broad absorption spectrum. 9-(1-Naphthyl)-10-(2-naphthyl)anthracene is luminescent, emitting both fluorescent and phosphorescent light. The material is fabricated using the Suzuki coupling reaction. Used in OLEDs as Fluorescent Host with superb lifetime. It can also be used as a surface active agent or surfactant in industrial applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unveiling the Root Cause of the Efficiency-Lifetime Trade-Off in Blue Fluorescent Organic Light-Emitting Diodes.
Sunwoo K, et al.
Electronic Materials Letters, 16(1), 1-8 (2020)
Superb lifetime of blue organic light-emitting diodes through engineering interface carrier blocking layers and adjusting electron leakage and an unusual efficiency variation at low electric field.
Taekyung K, et al.
Journal of Material Chemistry C, 6(31), 8472-8478 (2018)
Tsubasa Sasaki et al.
Nature communications, 12(1), 2706-2706 (2021-05-13)
Although significant progress has been made in the development of light-emitting materials for organic light-emitting diodes along with the elucidation of emission mechanisms, the electron injection/transport mechanism remains unclear, and the materials used for electron injection/transport have been basically unchanged

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service