Skip to Content
Merck
All Photos(3)

Documents

556696

Sigma-Aldrich

N,N′-Di(1-naphthyl)-N,N′-diphenyl-(1,1′-biphenyl)-4,4′-diamine

sublimed grade, 99%

Synonym(s):

N,N′-Bis(naphthalen-1-yl)-N,N′-bis(phenyl)benzidine, NPB, NPD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H32N2
CAS Number:
Molecular Weight:
588.74
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

sublimed grade

Assay

99%

loss

0.5 wt. % loss on heating, 345°C (typical, TGA)

mp

279-283 °C (lit.)

Orbital energy

HOMO 5.5 eV 
LUMO 2.4 eV 

OLED Device Performance

ITO/NPD/TCTA/BCPO:Ir(piq)3 (7-8%)/BCP/Alq3/LiF/Al

  • Color: red
  • Max. Luminance: 24529 Cd/m2
  • Max. EQE: 17 %
  • Turn-On Voltage: 2.7 V

ITO/NPD/TCTA/BCPO:Ir(ppy)3 (7-8%)/BCP/Alq3/LiF/Al
  • Color: green
  • Max. Luminance: 207839 Cd/m2
  • Max. EQE: 21.6 %
  • Turn-On Voltage: 2.1 V

ITO/NPD/mCP/BCPO:FIrpic (8%)/TAZ/LiF/Al
  • Color: blue
  • Max. Luminance: 35677 Cd/m2
  • Max. EQE: 23.5 %
  • Turn-On Voltage: 2.8 V

ITO/PEDOT:PSS/NPD/TCTA/m-CBTZ:Ir(Bt)2(acac) (10%)/TPBI/LiF/Al
  • Color: yellow
  • Max. Luminance: 106000 Cd/m2
  • Max. EQE: 17.5 %
  • Turn-On Voltage: 3 V

SMILES string

c1ccc(cc1)N(c2ccc(cc2)-c3ccc(cc3)N(c4ccccc4)c5cccc6ccccc56)c7cccc8ccccc78

InChI

1S/C44H32N2/c1-3-17-37(18-4-1)45(43-23-11-15-35-13-7-9-21-41(35)43)39-29-25-33(26-30-39)34-27-31-40(32-28-34)46(38-19-5-2-6-20-38)44-24-12-16-36-14-8-10-22-42(36)44/h1-32H

InChI key

IBHBKWKFFTZAHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TGA/DSC Lot specific traces available upon request

Application

Hole transporting material for OLED devices

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shakeel S Dalal et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(14), 4227-4232 (2015-04-02)
Physical vapor deposition is commonly used to prepare organic glasses that serve as the active layers in light-emitting diodes, photovoltaics, and other devices. Recent work has shown that orienting the molecules in such organic semiconductors can significantly enhance device performance.
Courtney M Wheatley et al.
Respiratory medicine, 109(8), 1001-1011 (2015-06-17)
Despite the demonstrated advantageous systemic changes in response to regular exercise for individuals with cystic fibrosis (CF), exercise is still viewed as an elective rather than a vital component of therapy, and it is likely that these benefits extend to
Highly efficient and stable organic light-emitting diode using 4,4'-bis(N-carbazolyl)-9,9'-spirobifluorene as a thermally stable host material
Tsuzuki, T.; Tokito, S.
Applied Physics Letters, 94, 033302-033302 (2009)
Phenylcarbazole-dipyridyl triazole hybrid as bipolar host material for phosphorescent OLEDs
Hung, W.; et al.
Journal of Materials Chemistry, 22, 5410-5418 (2012)
Davalyn R Powell et al.
Developmental biology, 407(2), 289-299 (2015-08-11)
Neural crest cells (NCCs) are essential embryonic progenitor cells that are unique to vertebrates and form a remarkably complex and coordinated system of highly motile cells. Migration of NCCs occurs along specific pathways within the embryo in response to both

Articles

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

Sublimed materials for organic electronic devices such of OFETs and OTFTs allow the achievement of better electronic properties, and may help increase a device’s lifetime.

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service