Skip to Content
Merck
All Photos(1)

Key Documents

926388

Sigma-Aldrich

HAT-CN

≥98% (sublimed)

Synonym(s):

Dipyrazino[2,3-f :2′,3′-h ]quinoxaline-2,3,6,7,10,11-hexacarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18N12
CAS Number:
Molecular Weight:
384.27
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Quality Level

Assay

≥98% (sublimed)

form

powder

band gap

3.9 eV

impurities

0.5% weight loss (TGA, > 400 ºC)

color

yellow

refractive index

n/550 1.81

transition temp

Tg 130 °C
Tm 265 °C

solubility

acetone: soluble
acetonitrile: soluble
methylene chloride: soluble

λmax

282-321 nm in methylene chloride (PL: 422 nm)

Orbital energy

HOMO -9.1 eV 
LUMO -5.2 eV 

semiconductor properties

(mobility=0.1-1 cm2/V·s) (electron mobility)

SMILES string

n1c2c(nc(c1C#N)C#N)c3nc(c(nc3c4nc(c(nc42)C#N)C#N)C#N)C#N

InChI

1S/C18N12/c19-1-7-8(2-20)26-14-13(25-7)15-17(29-10(4-22)9(3-21)27-15)18-16(14)28-11(5-23)12(6-24)30-18

InChI key

DKHNGUNXLDCATP-UHFFFAOYSA-N

Application

HAT-CN can be used in studies such as organic electronics, as a charge-transport material, electron-transport material, or active layer component. Its high electron affinity and π-conjugated structure suggest potential use in organic field-effect transistors (OFETs), organic photovoltaics (OPVs), and organic light-emitting diodes (OLEDs). HAT-CN may be studied for potential applications in energy storage, such as batteries and supercapacitors.
HAT-CN is a popular solution-processable hole transport material (HTM) for:

  • Diodes ;
  • Environmentally-friendly organic solar cells (OPVs);
  • Organic phototransistors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamic Oscillation via Negative Differential Resistance in Type III Junction Organic/Two-Dimensional and Oxide/Two-Dimensional Transition Metal Dichalcogenide Diodes.
Choi W, et al.
Advances in Functional Materials, 31(9), 2009436-2009436 (2021)
Effects on p-type buffer layers for semitransparent organic photovoltaics using indium zinc oxide transparent electrode.
Kuwano K, et al.
Japanese Journal of Applied Physics, 59(SD), SDDD04-SDDD04 (2020)
Solution-processed hexaazatriphenylene hexacarbonitrile as a universal hole-injection layer for organic light-emitting diodes
Lin H W, et al.
Organic Electronics, 14, 1204?1210-1204?1210 (2013)
Novel hole transporting materials based on 4-(9H-carbazol-9-yl)triphenylamine derivatives for OLEDs
Nguyen Q P B, et al.
Molecules (Basel), 19(9), 14247-14256 (2014)
Highly efficient green phosphorescent organic light-emitting diodes with low efficiency roll-off based on iridium(III) complexes bearing oxadiazol-substituted amide ligands.
Zhang F, et al.
Journal of Material Chemistry C, 4(23), 5469-5475 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service