Skip to Content
Merck
All Photos(1)

Key Documents

922773

Sigma-Aldrich

IPr*Pd(acac)Cl

≥95%

Synonym(s):

N,N′-Bis(2,6-bis(diphenylmethyl)-4-methylphenyl)imidazolium palladium acetylacetone chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C74H63ClN2O2Pd
CAS Number:
Molecular Weight:
1154.18
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or chunks

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

SMILES string

Cl[Pd]1(C2N(C(C(C(C3=CC=CC=C3)C4=CC=CC=C4)=C5)=C(C(C6=CC=CC=C6)C7=CC=CC=C7)C=C5C)C=CN2C(C(C(C8=CC=CC=C8)C9=CC=CC=C9)=C%10)=C(C(C%11=CC=CC=C%11)C%12=CC=CC=C%12)C=C%10C)oc(C)cc(C)o1

Application

IPr*Pd(acac)Cl is a palladium NHC precatalyst for a variety of cross-couplng reactions, including C-N bond formation.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Pd(IPr*)(acac)Cl]: An Easily Synthesized, Bulky Precatalyst for C?N Bond Formation
Meiries S, et al.
Organometallics, 31, 3402-3409 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service