Skip to Content
Merck
All Photos(3)

Documents

643777

Sigma-Aldrich

Di-tert-butyl(methyl)phosphonium tetrafluoroborate

97%

Synonym(s):

(t-Bu)2PMeHBF4, Bis(1,1-dimethylethyl)methylphosphine tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H22BF4P
CAS Number:
Molecular Weight:
248.05
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

pH

1.79 (1% in solution)

mp

>230 °C (lit.)

functional group

phosphine

SMILES string

F[B-](F)(F)F.C[PH+](C(C)(C)C)C(C)(C)C

InChI

1S/C9H21P.BF4/c1-8(2,3)10(7)9(4,5)6;2-1(3,4)5/h1-7H3;/q;-1/p+1

InChI key

BRDLRXCAHKUWJS-UHFFFAOYSA-O

Looking for similar products? Visit Product Comparison Guide

Application

Di-tert-butyl(methyl)phosphonium tetrafluoroborate may be used in the preparation of a precursor for nantenine analogs bearing a C4 phenyl substituent, which shows an affinity for 5-HT2B receptor. It may also be used in the palladium-catalyzed borylation of primary alkyl electrophiles (bromides, iodides or tosylates) using bis(pinacolato)diboron or bis(neopentyl glycolato)diboron as the boron source.
The phosphonium salt is used for Pd catalyzed cross coupling reaction.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-catalyzed borylation of primary alkyl bromides.
Joshi-Pangu A, et al.
The Journal of Organic Chemistry, 77(15), 6629-6633 (2012)
Laurence Caron et al.
Organic letters, 10(20), 4533-4536 (2008-09-25)
Direct arylation reactions of nitrobenzenes and aryl halides occur in good yield and high ortho regioselectivity. These reactions can be performed on gram scale with as few as 3 equiv of the nitro arene relative to the aryl halide. The
C4 phenyl aporphines with selective h5-HT 2B receptor affinity.
Kapadia N and Harding WW.
Bioorganic & Medicinal Chemistry Letters, 25(17), 3451-3454 (2015)
Jian-Xin Wang et al.
Organic letters, 10(14), 2923-2926 (2008-06-26)
A general and efficient synthesis of 5-aryl imidazo[1,5- a]pyrazines by palladium-catalyzed coupling of the corresponding 8-substituted derivatives with aryl halides is described. The scope of this new reaction for the imidazo[1,5- a]pyrazine ring system was explored using three readily available
Marc Lafrance et al.
Journal of the American Chemical Society, 128(27), 8754-8756 (2006-07-06)
Penta-, tetra-, tri-, and difluorobenzenes undergo direct arylation with a wide range of arylhalides in high yield. Inverse reactivity is observed compared to the common electrophilic aromatic substitution pathway since electron-deficient, C-H acidic arenes react preferentially. Computational studies indicate that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service